Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139536879-139537138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F9
Entrez Gene ID
2158
Gene Name
coagulation factor IX
Gene Aliases
F9 p22, FIX, HEMB, P19, PTC, THPH8
UniGene
Hs.522798
Gene Chromosome Location
Chr.X: 139530720-139563459 on Build GRCh38
Transcripts
NM_000133.3 NM_001313913.1 XM_005262397.4

SNP Detail

SNP ID Chromosome Location
rs200505648 Chr.X: 139537037-139537037 on Build GRCh38
rs184837275 Chr.X: 139537029-139537029 on Build GRCh38
rs764767654 Chr.X: 139536938-139536938 on Build GRCh38
rs757059944 Chr.X: 139536984-139536984 on Build GRCh38
rs759987427 Chr.X: 139537084-139537084 on Build GRCh38
rs757989062 Chr.X: 139536966-139536966 on Build GRCh38
rs920067525 Chr.X: 139536930-139536930 on Build GRCh38
rs367569299 Chr.X: 139537030-139537030 on Build GRCh38
rs928672798 Chr.X: 139537025-139537025 on Build GRCh38
rs745786675 Chr.X: 139536993-139536993 on Build GRCh38
rs756148155 Chr.X: 139537024-139537024 on Build GRCh38
rs137852223 Chr.X: 139537090-139537090 on Build GRCh38
rs751235513 Chr.X: 139536983-139536983 on Build GRCh38
rs139089559 Chr.X: 139537098-139537098 on Build GRCh38
rs745353370 Chr.X: 139537035-139537035 on Build GRCh38
rs779083411 Chr.X: 139537026-139537026 on Build GRCh38
rs749406722 Chr.X: 139536979-139536979 on Build GRCh38
rs756939161 Chr.X: 139536943-139536943 on Build GRCh38
rs763814160 Chr.X: 139536975-139536975 on Build GRCh38
rs774915238 Chr.X: 139537102-139537102 on Build GRCh38
rs776894974 Chr.X: 139537051-139537051 on Build GRCh38
rs780897125 Chr.X: 139536985-139536985 on Build GRCh38
rs371373268 Chr.X: 139537043-139537043 on Build GRCh38

View Full Product Details