Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27748060-27748291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8L2
Entrez Gene ID
347442
Gene Name
DDB1 and CUL4 associated factor 8 like 2
Gene Aliases
WDR42C
UniGene
Hs.720885
Gene Chromosome Location
Chr.X: 27746809-27748821 on Build GRCh38
Transcripts
NM_001136533.1

SNP Detail

SNP ID Chromosome Location
rs780562926 Chr.X: 27748222-27748222 on Build GRCh38
rs769551706 Chr.X: 27748256-27748256 on Build GRCh38
rs1011873595 Chr.X: 27748155-27748155 on Build GRCh38
rs976570296 Chr.X: 27748244-27748244 on Build GRCh38
rs368376110 Chr.X: 27748122-27748122 on Build GRCh38
rs750469981 Chr.X: 27748220-27748220 on Build GRCh38
rs761465771 Chr.X: 27748115-27748115 on Build GRCh38
rs1029038901 Chr.X: 27748249-27748249 on Build GRCh38
rs747363358 Chr.X: 27748103-27748103 on Build GRCh38
rs185264593 Chr.X: 27748253-27748253 on Build GRCh38
rs763034072 Chr.X: 27748138-27748138 on Build GRCh38
rs1056165880 Chr.X: 27748136-27748136 on Build GRCh38
rs909436093 Chr.X: 27748088-27748088 on Build GRCh38
rs61730883 Chr.X: 27748125-27748125 on Build GRCh38
rs754070602 Chr.X: 27748192-27748192 on Build GRCh38
rs766389302 Chr.X: 27748158-27748158 on Build GRCh38
rs966189526 Chr.X: 27748228-27748228 on Build GRCh38
rs779012547 Chr.X: 27748213-27748213 on Build GRCh38
rs767272705 Chr.X: 27748191-27748191 on Build GRCh38
rs769018767 Chr.X: 27748107-27748107 on Build GRCh38
rs759260989 Chr.X: 27748177-27748177 on Build GRCh38
rs751476265 Chr.X: 27748176-27748176 on Build GRCh38
rs757488857 Chr.X: 27748206-27748206 on Build GRCh38
rs781702266 Chr.X: 27748121-27748121 on Build GRCh38
rs868486185 Chr.X: 27748160-27748160 on Build GRCh38
rs747493517 Chr.X: 27748227-27748227 on Build GRCh38
rs776165375 Chr.X: 27748109-27748109 on Build GRCh38

View Full Product Details