Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 50590796-50591039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL1
Entrez Gene ID
51062
Gene Name
atlastin GTPase 1
Gene Aliases
AD-FSP, FSP1, GBP3, HSN1D, SPG3, SPG3A, atlastin1
UniGene
Hs.584905
Gene Chromosome Location
Chr.14: 50533082-50633068 on Build GRCh38
Transcripts
NM_001127713.1 NM_015915.4 NM_181598.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1369964 c.283G>A p.E95K Chr.14: 50590941-50590941 on Build GRCh38
3999275 c.306C>A p.D102E Chr.14: 50590964-50590964 on Build GRCh38
1707325 c.292G>A p.D98N Chr.14: 50590950-50590950 on Build GRCh38
1369965 c.352C>T p.R118* Chr.14: 50591010-50591010 on Build GRCh38
1607706 c.339A>T p.R113S Chr.14: 50590997-50590997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780987119 Chr.14: 50590895-50590895 on Build GRCh38
rs863224772 Chr.14: 50590958-50590958 on Build GRCh38
rs769529177 Chr.14: 50590898-50590898 on Build GRCh38
rs112496709 Chr.14: 50590980-50590980 on Build GRCh38
rs565002494 Chr.14: 50590862-50590862 on Build GRCh38
rs754379526 Chr.14: 50590926-50590926 on Build GRCh38
rs774586106 Chr.14: 50590900-50590900 on Build GRCh38
rs759686747 Chr.14: 50590904-50590904 on Build GRCh38
rs145204580 Chr.14: 50590964-50590964 on Build GRCh38
rs377736535 Chr.14: 50590969-50590969 on Build GRCh38
rs182560221 Chr.14: 50590871-50590871 on Build GRCh38
rs779559817 Chr.14: 50590998-50590998 on Build GRCh38
rs775694786 Chr.14: 50590917-50590917 on Build GRCh38
rs1060197 Chr.14: 50591009-50591009 on Build GRCh38
rs74051570 Chr.14: 50590885-50590885 on Build GRCh38
rs747986329 Chr.14: 50590897-50590897 on Build GRCh38
rs765257350 Chr.14: 50590853-50590853 on Build GRCh38
rs905849725 Chr.14: 50590872-50590872 on Build GRCh38
rs544975285 Chr.14: 50590850-50590850 on Build GRCh38
rs1048820808 Chr.14: 50590849-50590849 on Build GRCh38
rs754423972 Chr.14: 50591010-50591010 on Build GRCh38
rs760663394 Chr.14: 50590920-50590920 on Build GRCh38
rs764697428 Chr.14: 50590925-50590925 on Build GRCh38
rs606231265 Chr.14: 50591011-50591011 on Build GRCh38
rs753620247 Chr.14: 50590833-50590833 on Build GRCh38

View Full Product Details