Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73816821-73817046 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf78
Entrez Gene ID
388960
Gene Name
chromosome 2 open reading frame 78
Gene Aliases
COG5373, hCG1989538
UniGene
Hs.456833
Gene Chromosome Location
Chr.2: 73784189-73817148 on Build GRCh38
Transcripts
NM_001080474.1 XM_005264302.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7170957 c.2542G>A p.E848K Chr.2: 73816855-73816855 on Build GRCh38
4837934 c.2646G>T p.M882I Chr.2: 73816959-73816959 on Build GRCh38
5032090 c.2636G>A p.R879H Chr.2: 73816859-73816859 on Build GRCh38
4834454 c.2656G>C p.E886Q Chr.2: 73816969-73816969 on Build GRCh38
5032091 c.2546G>A p.R849H Chr.2: 73816859-73816859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549195445 Chr.2: 73816968-73816968 on Build GRCh38
rs887643021 Chr.2: 73816961-73816961 on Build GRCh38
rs758443703 Chr.2: 73816852-73816852 on Build GRCh38
rs751614565 Chr.2: 73816985-73816985 on Build GRCh38
rs766473401 Chr.2: 73816959-73816959 on Build GRCh38
rs35648065 Chr.2: 73816960-73816961 on Build GRCh38
rs765378248 Chr.2: 73816983-73816983 on Build GRCh38
rs762272994 Chr.2: 73816940-73816940 on Build GRCh38
rs562767008 Chr.2: 73816887-73816887 on Build GRCh38
rs756025346 Chr.2: 73816874-73816874 on Build GRCh38
rs781056663 Chr.2: 73817007-73817007 on Build GRCh38
rs267599452 Chr.2: 73816960-73816960 on Build GRCh38
rs748756915 Chr.2: 73816892-73816892 on Build GRCh38
rs774884448 Chr.2: 73816928-73816928 on Build GRCh38
rs373266021 Chr.2: 73816859-73816859 on Build GRCh38
rs769173412 Chr.2: 73816927-73816927 on Build GRCh38
rs369360208 Chr.2: 73816858-73816858 on Build GRCh38
rs947815971 Chr.2: 73816891-73816891 on Build GRCh38
rs962816078 Chr.2: 73816948-73816948 on Build GRCh38
rs370246642 Chr.2: 73816958-73816958 on Build GRCh38
rs758442837 Chr.2: 73816975-73816975 on Build GRCh38
rs376212025 Chr.2: 73816879-73816879 on Build GRCh38
rs865779725 Chr.2: 73816975-73816975 on Build GRCh38
rs916294284 Chr.2: 73816890-73816890 on Build GRCh38
rs773433099 Chr.2: 73816951-73816951 on Build GRCh38
rs757130074 Chr.2: 73816989-73816989 on Build GRCh38
rs1028176434 Chr.2: 73816998-73816998 on Build GRCh38
rs767988843 Chr.2: 73816947-73816947 on Build GRCh38
rs201654918 Chr.2: 73816857-73816857 on Build GRCh38

View Full Product Details