Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52587036-52587265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM214A
Entrez Gene ID
56204
Gene Name
family with sequence similarity 214 member A
Gene Aliases
KIAA1370
UniGene
Hs.152385
Gene Chromosome Location
Chr.15: 52581321-52709817 on Build GRCh38
Transcripts
NM_001286495.1 NM_019600.3 NR_104457.1 XM_005254547.2 XM_005254548.2 XM_011521789.2 XM_011521790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
673579 c.2809-10T>C p.? Chr.15: 52587229-52587229 on Build GRCh38
7165812 c.2861C>T p.S954L Chr.15: 52587167-52587167 on Build GRCh38
1478192 c.2858C>T p.P953L Chr.15: 52587170-52587170 on Build GRCh38
4931870 c.2877+1G>A p.? Chr.15: 52587150-52587150 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762646762 Chr.15: 52587100-52587100 on Build GRCh38
rs371037711 Chr.15: 52587194-52587194 on Build GRCh38
rs765731613 Chr.15: 52587089-52587089 on Build GRCh38
rs772695177 Chr.15: 52587095-52587095 on Build GRCh38
rs369682402 Chr.15: 52587166-52587166 on Build GRCh38
rs777521181 Chr.15: 52587178-52587178 on Build GRCh38
rs765989672 Chr.15: 52587101-52587101 on Build GRCh38
rs754542048 Chr.15: 52587132-52587132 on Build GRCh38
rs755853433 Chr.15: 52587171-52587171 on Build GRCh38
rs767385911 Chr.15: 52587139-52587139 on Build GRCh38
rs772601984 Chr.15: 52587230-52587232 on Build GRCh38
rs749076396 Chr.15: 52587191-52587191 on Build GRCh38
rs751077589 Chr.15: 52587103-52587103 on Build GRCh38

View Full Product Details