Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 60005804-60006061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXB1
Entrez Gene ID
27023
Gene Name
forkhead box B1
Gene Aliases
FKH5, HFKH-5
UniGene
Hs.734475
Gene Chromosome Location
Chr.15: 60004222-60005943 on Build GRCh38
Transcripts
NM_012182.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6409090 c.907A>C p.S303R Chr.15: 60005870-60005870 on Build GRCh38
4955137 c.949T>C p.S317P Chr.15: 60005912-60005912 on Build GRCh38
700817 c.846C>A p.T282T Chr.15: 60005809-60005809 on Build GRCh38
700816 c.918C>A p.A306A Chr.15: 60005881-60005881 on Build GRCh38
4756198 c.968C>T p.A323V Chr.15: 60005931-60005931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747097444 Chr.15: 60005858-60005858 on Build GRCh38
rs780492307 Chr.15: 60005857-60005857 on Build GRCh38
rs750900043 Chr.15: 60005849-60005849 on Build GRCh38
rs750808428 Chr.15: 60005961-60005961 on Build GRCh38
rs757668305 Chr.15: 60005844-60005844 on Build GRCh38
rs981253679 Chr.15: 60005971-60005971 on Build GRCh38
rs746281359 Chr.15: 60005924-60005924 on Build GRCh38
rs756335501 Chr.15: 60005976-60005976 on Build GRCh38
rs758975065 Chr.15: 60005851-60005851 on Build GRCh38
rs774317103 Chr.15: 60005987-60005987 on Build GRCh38
rs188651499 Chr.15: 60005980-60005980 on Build GRCh38
rs759067888 Chr.15: 60005920-60005920 on Build GRCh38
rs774216321 Chr.15: 60005917-60005917 on Build GRCh38
rs376335632 Chr.15: 60005962-60005962 on Build GRCh38
rs760413711 Chr.15: 60005945-60005945 on Build GRCh38
rs374565718 Chr.15: 60005847-60005847 on Build GRCh38
rs781156396 Chr.15: 60005972-60005972 on Build GRCh38
rs773004677 Chr.15: 60005904-60005904 on Build GRCh38
rs748313472 Chr.15: 60005974-60005974 on Build GRCh38
rs748401698 Chr.15: 60005866-60005866 on Build GRCh38
rs767188993 Chr.15: 60005825-60005825 on Build GRCh38
rs72739945 Chr.15: 60005966-60005966 on Build GRCh38
rs745826978 Chr.15: 60005991-60005991 on Build GRCh38
rs546991668 Chr.15: 60005862-60005862 on Build GRCh38
rs765627268 Chr.15: 60005952-60005952 on Build GRCh38
rs887806872 Chr.15: 60005914-60005914 on Build GRCh38
rs769531193 Chr.15: 60005871-60005871 on Build GRCh38
rs749156821 Chr.15: 60005910-60005910 on Build GRCh38
rs777629396 Chr.15: 60005983-60005983 on Build GRCh38
rs139129563 Chr.15: 60005890-60005890 on Build GRCh38
rs768665651 Chr.15: 60005859-60005859 on Build GRCh38
rs918910291 Chr.15: 60005978-60005978 on Build GRCh38
rs770685026 Chr.15: 60005986-60005986 on Build GRCh38
rs752344376 Chr.15: 60005833-60005833 on Build GRCh38
rs770854457 Chr.15: 60005915-60005915 on Build GRCh38
rs539373129 Chr.15: 60005934-60005934 on Build GRCh38
rs991735501 Chr.15: 60005959-60005959 on Build GRCh38
rs8040887 Chr.15: 60006022-60006022 on Build GRCh38
rs775156922 Chr.15: 60005936-60005936 on Build GRCh38
rs749073690 Chr.15: 60005985-60005985 on Build GRCh38
rs957170682 Chr.15: 60005932-60005932 on Build GRCh38

View Full Product Details