Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8477269-8477536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC6A
Entrez Gene ID
93978
Gene Name
C-type lectin domain family 6 member A
Gene Aliases
CLEC4N, CLECSF10
UniGene
Hs.591147
Gene Chromosome Location
Chr.12: 8455995-8478330 on Build GRCh38
Transcripts
NM_001007033.1 NM_001317999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
944152 c.488T>G p.F163C Chr.12: 8477322-8477322 on Build GRCh38
3466162 c.515C>T p.S172F Chr.12: 8477349-8477349 on Build GRCh38
3466163 c.556G>A p.G186R Chr.12: 8477390-8477390 on Build GRCh38
5084309 c.492G>A p.W164* Chr.12: 8477326-8477326 on Build GRCh38
3466164 c.594G>A p.R198R Chr.12: 8477428-8477428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759811422 Chr.12: 8477415-8477415 on Build GRCh38
rs752704093 Chr.12: 8477313-8477313 on Build GRCh38
rs759471781 Chr.12: 8477303-8477303 on Build GRCh38
rs199626477 Chr.12: 8477489-8477489 on Build GRCh38
rs756160870 Chr.12: 8477318-8477318 on Build GRCh38
rs771849227 Chr.12: 8477364-8477364 on Build GRCh38
rs768095545 Chr.12: 8477470-8477470 on Build GRCh38
rs773611047 Chr.12: 8477403-8477403 on Build GRCh38
rs759897542 Chr.12: 8477503-8477503 on Build GRCh38
rs111454531 Chr.12: 8477463-8477463 on Build GRCh38
rs778920033 Chr.12: 8477448-8477448 on Build GRCh38
rs766767003 Chr.12: 8477408-8477408 on Build GRCh38
rs746636942 Chr.12: 8477465-8477465 on Build GRCh38
rs755158438 Chr.12: 8477307-8477307 on Build GRCh38
rs901401938 Chr.12: 8477393-8477393 on Build GRCh38
rs745648028 Chr.12: 8477449-8477449 on Build GRCh38
rs781314503 Chr.12: 8477309-8477309 on Build GRCh38
rs774690418 Chr.12: 8477412-8477412 on Build GRCh38
rs749210704 Chr.12: 8477338-8477338 on Build GRCh38
rs758212284 Chr.12: 8477457-8477457 on Build GRCh38
rs772355833 Chr.12: 8477488-8477490 on Build GRCh38
rs763490309 Chr.12: 8477407-8477407 on Build GRCh38
rs141483310 Chr.12: 8477361-8477361 on Build GRCh38
rs771412426 Chr.12: 8477500-8477500 on Build GRCh38
rs766677073 Chr.12: 8477295-8477295 on Build GRCh38
rs111292684 Chr.12: 8477502-8477502 on Build GRCh38
rs149856067 Chr.12: 8477369-8477369 on Build GRCh38
rs751825764 Chr.12: 8477298-8477298 on Build GRCh38
rs780920394 Chr.12: 8477349-8477349 on Build GRCh38
rs753768710 Chr.12: 8477438-8477438 on Build GRCh38
rs775845223 Chr.12: 8477506-8477507 on Build GRCh38
rs776171426 Chr.12: 8477477-8477477 on Build GRCh38
rs746519277 Chr.12: 8477372-8477372 on Build GRCh38
rs140087240 Chr.12: 8477422-8477422 on Build GRCh38
rs757231031 Chr.12: 8477444-8477444 on Build GRCh38
rs1004349180 Chr.12: 8477320-8477320 on Build GRCh38
rs866329197 Chr.12: 8477379-8477379 on Build GRCh38
rs777575034 Chr.12: 8477329-8477329 on Build GRCh38
rs745644404 Chr.12: 8477398-8477398 on Build GRCh38
rs764185220 Chr.12: 8477437-8477437 on Build GRCh38
rs762340299 Chr.12: 8477302-8477302 on Build GRCh38
rs779580365 Chr.12: 8477461-8477461 on Build GRCh38
rs756178091 Chr.12: 8477434-8477434 on Build GRCh38
rs114953954 Chr.12: 8477433-8477433 on Build GRCh38
rs778830183 Chr.12: 8477360-8477360 on Build GRCh38
rs900589102 Chr.12: 8477490-8477490 on Build GRCh38
rs757140928 Chr.12: 8477341-8477341 on Build GRCh38
rs566165884 Chr.12: 8477392-8477392 on Build GRCh38
rs147949525 Chr.12: 8477397-8477397 on Build GRCh38

View Full Product Details