Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2749236-2749465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBF4
Entrez Gene ID
57593
Gene Name
early B-cell factor 4
Gene Aliases
COE4, O/E-4
UniGene
Hs.471955
Gene Chromosome Location
Chr.20: 2692878-2760108 on Build GRCh38
Transcripts
NM_001110514.1 XM_006723600.1 XM_017027983.1 XM_017027984.1 XM_017027985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504915 c.678C>T p.S226S Chr.20: 2749451-2749451 on Build GRCh38
5554939 c.658G>A p.G220R Chr.20: 2749431-2749431 on Build GRCh38
3693413 c.679G>A p.D227N Chr.20: 2749452-2749452 on Build GRCh38
4410192 c.657C>T p.D219D Chr.20: 2749430-2749430 on Build GRCh38
4624221 c.644C>T p.T215M Chr.20: 2749417-2749417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747038739 Chr.20: 2749300-2749300 on Build GRCh38
rs953811128 Chr.20: 2749413-2749413 on Build GRCh38
rs1011045039 Chr.20: 2749322-2749322 on Build GRCh38
rs547059866 Chr.20: 2749310-2749310 on Build GRCh38
rs554847712 Chr.20: 2749445-2749445 on Build GRCh38
rs186518884 Chr.20: 2749393-2749393 on Build GRCh38
rs1029315262 Chr.20: 2749406-2749406 on Build GRCh38
rs966447063 Chr.20: 2749386-2749386 on Build GRCh38
rs911873478 Chr.20: 2749439-2749439 on Build GRCh38
rs989788108 Chr.20: 2749347-2749347 on Build GRCh38
rs764057057 Chr.20: 2749370-2749370 on Build GRCh38
rs1034325329 Chr.20: 2749297-2749297 on Build GRCh38
rs566006616 Chr.20: 2749363-2749363 on Build GRCh38
rs775322269 Chr.20: 2749362-2749362 on Build GRCh38
rs1002473245 Chr.20: 2749260-2749260 on Build GRCh38
rs985411829 Chr.20: 2749436-2749436 on Build GRCh38
rs958234777 Chr.20: 2749307-2749307 on Build GRCh38
rs773251916 Chr.20: 2749380-2749380 on Build GRCh38
rs978100825 Chr.20: 2749415-2749415 on Build GRCh38
rs1033854061 Chr.20: 2749334-2749334 on Build GRCh38
rs766473303 Chr.20: 2749428-2749428 on Build GRCh38
rs538592350 Chr.20: 2749257-2749257 on Build GRCh38
rs959771563 Chr.20: 2749345-2749345 on Build GRCh38

View Full Product Details