Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45806927-45807194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR1
Entrez Gene ID
1394
Gene Name
corticotropin releasing hormone receptor 1
Gene Aliases
CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L
UniGene
Hs.417628
Gene Chromosome Location
Chr.17: 45784280-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Gene Symbol
MGC57346-CRHR1
Entrez Gene ID
104909134
Gene Name
MGC57346-CRHR1 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 45620329-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612468 c.79G>A p.D27N Chr.17: 45807055-45807055 on Build GRCh38
7185348 c.97C>T p.L33L Chr.17: 45807073-45807073 on Build GRCh38
5890963 c.121+8C>T p.? Chr.17: 45807105-45807105 on Build GRCh38
4943074 c.68C>T p.A23V Chr.17: 45807044-45807044 on Build GRCh38
359104 c.42C>A p.L14L Chr.17: 45807018-45807018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774998390 Chr.17: 45807037-45807037 on Build GRCh38
rs375401431 Chr.17: 45806972-45806972 on Build GRCh38
rs770517761 Chr.17: 45807026-45807026 on Build GRCh38
rs1056171833 Chr.17: 45807137-45807137 on Build GRCh38
rs543785844 Chr.17: 45807066-45807066 on Build GRCh38
rs12936511 Chr.17: 45807036-45807036 on Build GRCh38
rs532830576 Chr.17: 45807156-45807156 on Build GRCh38
rs758350424 Chr.17: 45807077-45807077 on Build GRCh38
rs755097274 Chr.17: 45806974-45806974 on Build GRCh38
rs765305113 Chr.17: 45807071-45807071 on Build GRCh38
rs780696864 Chr.17: 45807119-45807119 on Build GRCh38
rs748617125 Chr.17: 45807141-45807141 on Build GRCh38
rs746620826 Chr.17: 45806997-45806997 on Build GRCh38
rs774017748 Chr.17: 45806959-45806959 on Build GRCh38
rs777438253 Chr.17: 45806993-45806993 on Build GRCh38
rs757127263 Chr.17: 45807114-45807114 on Build GRCh38
rs905052602 Chr.17: 45807064-45807064 on Build GRCh38
rs369514071 Chr.17: 45807048-45807048 on Build GRCh38
rs766487218 Chr.17: 45807056-45807056 on Build GRCh38
rs763906366 Chr.17: 45807084-45807084 on Build GRCh38
rs779724823 Chr.17: 45807139-45807139 on Build GRCh38
rs753859538 Chr.17: 45807031-45807031 on Build GRCh38
rs372100277 Chr.17: 45807012-45807012 on Build GRCh38
rs745351116 Chr.17: 45807034-45807034 on Build GRCh38
rs1009680275 Chr.17: 45807153-45807153 on Build GRCh38
rs373210936 Chr.17: 45807146-45807146 on Build GRCh38
rs966094389 Chr.17: 45806954-45806954 on Build GRCh38
rs758119565 Chr.17: 45806992-45806992 on Build GRCh38
rs1017788119 Chr.17: 45806950-45806950 on Build GRCh38
rs752569982 Chr.17: 45807075-45807075 on Build GRCh38
rs755621596 Chr.17: 45807130-45807130 on Build GRCh38
rs757069588 Chr.17: 45807047-45807047 on Build GRCh38
rs937496116 Chr.17: 45807120-45807120 on Build GRCh38
rs780940635 Chr.17: 45807032-45807032 on Build GRCh38
rs772507840 Chr.17: 45807043-45807043 on Build GRCh38
rs759607266 Chr.17: 45807067-45807067 on Build GRCh38
rs368983783 Chr.17: 45807126-45807126 on Build GRCh38
rs112027273 Chr.17: 45807001-45807001 on Build GRCh38
rs199783024 Chr.17: 45807106-45807106 on Build GRCh38
rs1030373486 Chr.17: 45806965-45806965 on Build GRCh38
rs765059247 Chr.17: 45807117-45807117 on Build GRCh38
rs377696758 Chr.17: 45807111-45807111 on Build GRCh38
rs752697095 Chr.17: 45806986-45806986 on Build GRCh38
rs954847384 Chr.17: 45807007-45807007 on Build GRCh38
rs369389731 Chr.17: 45806981-45806981 on Build GRCh38

View Full Product Details