Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100805434-100805687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB4
Entrez Gene ID
2050
Gene Name
EPH receptor B4
Gene Aliases
HTK, MYK1, TYRO11
UniGene
Hs.437008
Gene Chromosome Location
Chr.7: 100802565-100827521 on Build GRCh38
Transcripts
NM_004444.4 XM_017011816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4992805 c.2605C>T p.Q869* Chr.7: 100805574-100805574 on Build GRCh38
6436372 c.2644G>A p.A882T Chr.7: 100805535-100805535 on Build GRCh38
4992806 c.2604C>T p.P868P Chr.7: 100805575-100805575 on Build GRCh38
20612 c.2665C>T p.R889W Chr.7: 100805514-100805514 on Build GRCh38
1205522 c.2597G>A p.R866H Chr.7: 100805582-100805582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149061889 Chr.7: 100805521-100805521 on Build GRCh38
rs146445434 Chr.7: 100805513-100805513 on Build GRCh38
rs953048390 Chr.7: 100805608-100805608 on Build GRCh38
rs752151153 Chr.7: 100805540-100805540 on Build GRCh38
rs763645511 Chr.7: 100805657-100805657 on Build GRCh38
rs752220598 Chr.7: 100805490-100805490 on Build GRCh38
rs34918225 Chr.7: 100805535-100805535 on Build GRCh38
rs767345190 Chr.7: 100805650-100805650 on Build GRCh38
rs763548237 Chr.7: 100805561-100805561 on Build GRCh38
rs984407128 Chr.7: 100805631-100805631 on Build GRCh38
rs755595176 Chr.7: 100805541-100805541 on Build GRCh38
rs372777956 Chr.7: 100805468-100805468 on Build GRCh38
rs780669240 Chr.7: 100805486-100805486 on Build GRCh38
rs781770682 Chr.7: 100805578-100805578 on Build GRCh38
rs201842419 Chr.7: 100805562-100805562 on Build GRCh38
rs146768192 Chr.7: 100805501-100805501 on Build GRCh38
rs759414784 Chr.7: 100805536-100805536 on Build GRCh38
rs549742543 Chr.7: 100805582-100805582 on Build GRCh38
rs774132008 Chr.7: 100805616-100805616 on Build GRCh38
rs375504500 Chr.7: 100805473-100805473 on Build GRCh38
rs747337230 Chr.7: 100805574-100805574 on Build GRCh38
rs756917361 Chr.7: 100805563-100805563 on Build GRCh38
rs751747096 Chr.7: 100805661-100805662 on Build GRCh38
rs762016655 Chr.7: 100805514-100805514 on Build GRCh38
rs369484009 Chr.7: 100805480-100805480 on Build GRCh38
rs766137248 Chr.7: 100805478-100805478 on Build GRCh38
rs755503621 Chr.7: 100805575-100805575 on Build GRCh38
rs975948410 Chr.7: 100805550-100805550 on Build GRCh38
rs770095206 Chr.7: 100805520-100805520 on Build GRCh38
rs769965440 Chr.7: 100805589-100805589 on Build GRCh38
rs753299011 Chr.7: 100805661-100805661 on Build GRCh38
rs35638378 Chr.7: 100805509-100805509 on Build GRCh38
rs776907382 Chr.7: 100805456-100805456 on Build GRCh38
rs771075650 Chr.7: 100805605-100805605 on Build GRCh38
rs750146958 Chr.7: 100805471-100805471 on Build GRCh38
rs375175851 Chr.7: 100805591-100805591 on Build GRCh38
rs763444841 Chr.7: 100805465-100805465 on Build GRCh38
rs747538171 Chr.7: 100805506-100805506 on Build GRCh38
rs774491104 Chr.7: 100805583-100805583 on Build GRCh38
rs763126310 Chr.7: 100805532-100805532 on Build GRCh38
rs140135838 Chr.7: 100805664-100805664 on Build GRCh38
rs775373372 Chr.7: 100805656-100805656 on Build GRCh38
rs753065467 Chr.7: 100805456-100805456 on Build GRCh38
rs777424003 Chr.7: 100805499-100805499 on Build GRCh38
rs754980362 Chr.7: 100805481-100805481 on Build GRCh38
rs767470095 Chr.7: 100805539-100805539 on Build GRCh38
rs140332222 Chr.7: 100805502-100805502 on Build GRCh38
rs373047132 Chr.7: 100805491-100805491 on Build GRCh38
rs148193109 Chr.7: 100805590-100805590 on Build GRCh38
rs764350075 Chr.7: 100805548-100805548 on Build GRCh38
rs765562710 Chr.7: 100805469-100805469 on Build GRCh38

View Full Product Details