Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47503347-47503582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
UniGene
Hs.485518
Gene Chromosome Location
Chr.6: 47477746-47627263 on Build GRCh38
Transcripts
NM_012120.2 XM_005248976.1 XM_017010641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153440 c.139G>A p.G47R Chr.6: 47503414-47503414 on Build GRCh38
6056625 c.165G>T p.K55N Chr.6: 47503440-47503440 on Build GRCh38
1319083 c.137G>T p.R46I Chr.6: 47503412-47503412 on Build GRCh38
3874582 c.86T>C p.V29A Chr.6: 47503361-47503361 on Build GRCh38
5068085 c.165+5G>T p.? Chr.6: 47503445-47503445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772360705 Chr.6: 47503382-47503382 on Build GRCh38
rs764144193 Chr.6: 47503435-47503435 on Build GRCh38
rs1025098433 Chr.6: 47503472-47503472 on Build GRCh38
rs368408367 Chr.6: 47503453-47503453 on Build GRCh38
rs983187870 Chr.6: 47503535-47503535 on Build GRCh38
rs758683074 Chr.6: 47503485-47503485 on Build GRCh38
rs1032565761 Chr.6: 47503438-47503438 on Build GRCh38
rs765069980 Chr.6: 47503420-47503421 on Build GRCh38
rs767203625 Chr.6: 47503428-47503428 on Build GRCh38
rs1001102537 Chr.6: 47503430-47503430 on Build GRCh38
rs375080675 Chr.6: 47503447-47503447 on Build GRCh38
rs780521436 Chr.6: 47503400-47503400 on Build GRCh38
rs770594307 Chr.6: 47503417-47503417 on Build GRCh38
rs759274707 Chr.6: 47503423-47503423 on Build GRCh38
rs935991276 Chr.6: 47503506-47503506 on Build GRCh38
rs796978855 Chr.6: 47503508-47503508 on Build GRCh38
rs372563870 Chr.6: 47503457-47503457 on Build GRCh38
rs772572598 Chr.6: 47503414-47503414 on Build GRCh38
rs970450708 Chr.6: 47503474-47503474 on Build GRCh38
rs551729252 Chr.6: 47503434-47503434 on Build GRCh38
rs151118470 Chr.6: 47503439-47503439 on Build GRCh38
rs370673360 Chr.6: 47503418-47503418 on Build GRCh38
rs762501100 Chr.6: 47503415-47503415 on Build GRCh38
rs747463684 Chr.6: 47503401-47503401 on Build GRCh38
rs759554468 Chr.6: 47503419-47503420 on Build GRCh38
rs769243431 Chr.6: 47503404-47503404 on Build GRCh38

View Full Product Details