Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96268424-96268697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIAO1
Entrez Gene ID
9391
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
CIA1, WDR39
UniGene
Hs.12109
Gene Chromosome Location
Chr.2: 96266146-96274179 on Build GRCh38
Transcripts
NM_004804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4987908 c.520G>A p.V174M Chr.2: 96268487-96268487 on Build GRCh38
6629158 c.601G>A p.A201T Chr.2: 96268568-96268568 on Build GRCh38
5964117 c.612G>A p.P204P Chr.2: 96268579-96268579 on Build GRCh38
3840279 c.511G>C p.D171H Chr.2: 96268478-96268478 on Build GRCh38
5005931 c.533G>A p.R178Q Chr.2: 96268500-96268500 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559101787 Chr.2: 96268552-96268552 on Build GRCh38
rs571238881 Chr.2: 96268604-96268604 on Build GRCh38
rs746325594 Chr.2: 96268645-96268645 on Build GRCh38
rs768342930 Chr.2: 96268668-96268668 on Build GRCh38
rs779467939 Chr.2: 96268475-96268475 on Build GRCh38
rs992845067 Chr.2: 96268524-96268524 on Build GRCh38
rs746419754 Chr.2: 96268529-96268529 on Build GRCh38
rs904753769 Chr.2: 96268583-96268583 on Build GRCh38
rs757668297 Chr.2: 96268637-96268637 on Build GRCh38
rs192314683 Chr.2: 96268597-96268597 on Build GRCh38
rs749741492 Chr.2: 96268490-96268491 on Build GRCh38
rs939231845 Chr.2: 96268548-96268548 on Build GRCh38
rs902409423 Chr.2: 96268544-96268544 on Build GRCh38
rs771485213 Chr.2: 96268520-96268520 on Build GRCh38
rs142522111 Chr.2: 96268614-96268614 on Build GRCh38
rs946021559 Chr.2: 96268537-96268537 on Build GRCh38
rs747374312 Chr.2: 96268504-96268504 on Build GRCh38
rs761581891 Chr.2: 96268590-96268590 on Build GRCh38
rs377203527 Chr.2: 96268500-96268500 on Build GRCh38
rs774622752 Chr.2: 96268589-96268589 on Build GRCh38
rs371292183 Chr.2: 96268632-96268632 on Build GRCh38
rs140260270 Chr.2: 96268650-96268650 on Build GRCh38
rs775568417 Chr.2: 96268664-96268664 on Build GRCh38
rs144987111 Chr.2: 96268579-96268579 on Build GRCh38
rs542568006 Chr.2: 96268452-96268452 on Build GRCh38
rs1004474331 Chr.2: 96268631-96268631 on Build GRCh38
rs749319878 Chr.2: 96268667-96268667 on Build GRCh38
rs199846474 Chr.2: 96268578-96268578 on Build GRCh38
rs199831944 Chr.2: 96268565-96268565 on Build GRCh38
rs1042341646 Chr.2: 96268531-96268531 on Build GRCh38
rs368621207 Chr.2: 96268527-96268527 on Build GRCh38
rs756030018 Chr.2: 96268603-96268603 on Build GRCh38
rs1027780080 Chr.2: 96268612-96268612 on Build GRCh38
rs184147889 Chr.2: 96268639-96268639 on Build GRCh38
rs754584475 Chr.2: 96268613-96268613 on Build GRCh38
rs936196574 Chr.2: 96268609-96268609 on Build GRCh38
rs767347306 Chr.2: 96268596-96268596 on Build GRCh38
rs771628407 Chr.2: 96268659-96268660 on Build GRCh38
rs1019316162 Chr.2: 96268652-96268652 on Build GRCh38
rs754601899 Chr.2: 96268498-96268498 on Build GRCh38
rs748924142 Chr.2: 96268480-96268480 on Build GRCh38
rs747856302 Chr.2: 96268622-96268622 on Build GRCh38
rs753292815 Chr.2: 96268607-96268607 on Build GRCh38
rs769876006 Chr.2: 96268530-96268530 on Build GRCh38

View Full Product Details