Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44148796-44149055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf53
Entrez Gene ID
78995
Gene Name
chromosome 17 open reading frame 53
Gene Aliases
-
UniGene
Hs.437059
Gene Chromosome Location
Chr.17: 44141902-44162477 on Build GRCh38
Transcripts
NM_001171251.2 NM_001321310.1 NM_001321311.1 NM_024032.4 XM_011525187.1 XM_011525188.1 XM_011525189.1 XM_011525190.1 XM_011525191.2 XM_011525192.1 XM_011525193.1 XM_011525194.1 XM_011525195.1 XM_017025031.1 XM_017025032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5664630 c.1220A>T p.H407L Chr.17: 44149023-44149023 on Build GRCh38
979886 c.1077C>A p.P359P Chr.17: 44148880-44148880 on Build GRCh38
5899315 c.1054C>T p.P352S Chr.17: 44148857-44148857 on Build GRCh38
4519751 c.1026G>T p.P342P Chr.17: 44148829-44148829 on Build GRCh38
5899314 c.1055C>T p.P352L Chr.17: 44148858-44148858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551307440 Chr.17: 44148913-44148913 on Build GRCh38
rs754276088 Chr.17: 44148833-44148833 on Build GRCh38
rs377054059 Chr.17: 44148884-44148884 on Build GRCh38
rs867913636 Chr.17: 44148857-44148857 on Build GRCh38
rs927825878 Chr.17: 44148849-44148849 on Build GRCh38
rs765714281 Chr.17: 44148952-44148952 on Build GRCh38
rs754379355 Chr.17: 44148990-44148990 on Build GRCh38
rs779915838 Chr.17: 44148987-44148987 on Build GRCh38
rs376112719 Chr.17: 44148902-44148902 on Build GRCh38
rs767384548 Chr.17: 44148974-44148974 on Build GRCh38
rs752761340 Chr.17: 44148855-44148855 on Build GRCh38
rs771236546 Chr.17: 44148944-44148944 on Build GRCh38
rs139569104 Chr.17: 44148828-44148828 on Build GRCh38
rs773942001 Chr.17: 44148914-44148914 on Build GRCh38
rs749106744 Chr.17: 44148837-44148837 on Build GRCh38
rs148903176 Chr.17: 44148978-44148978 on Build GRCh38
rs143555103 Chr.17: 44149010-44149010 on Build GRCh38
rs760058867 Chr.17: 44148839-44148839 on Build GRCh38
rs373604455 Chr.17: 44148945-44148945 on Build GRCh38
rs370350455 Chr.17: 44148899-44148899 on Build GRCh38
rs760007558 Chr.17: 44148948-44148948 on Build GRCh38
rs748828144 Chr.17: 44148989-44148989 on Build GRCh38
rs370848247 Chr.17: 44148905-44148905 on Build GRCh38
rs761458669 Chr.17: 44148943-44148943 on Build GRCh38
rs1035037095 Chr.17: 44148962-44148962 on Build GRCh38
rs201288595 Chr.17: 44148826-44148826 on Build GRCh38
rs769879941 Chr.17: 44148887-44148887 on Build GRCh38
rs750286909 Chr.17: 44148977-44148977 on Build GRCh38
rs946609560 Chr.17: 44149018-44149018 on Build GRCh38
rs145594792 Chr.17: 44148965-44148965 on Build GRCh38
rs934941110 Chr.17: 44148982-44148982 on Build GRCh38
rs757049747 Chr.17: 44148878-44148878 on Build GRCh38
rs990763800 Chr.17: 44148993-44148993 on Build GRCh38
rs778498532 Chr.17: 44148992-44148992 on Build GRCh38
rs763012183 Chr.17: 44148961-44148961 on Build GRCh38
rs761084854 Chr.17: 44148829-44148829 on Build GRCh38
rs777918652 Chr.17: 44148860-44148860 on Build GRCh38
rs367741443 Chr.17: 44148970-44148970 on Build GRCh38
rs372568603 Chr.17: 44148859-44148859 on Build GRCh38
rs759254344 Chr.17: 44148931-44148932 on Build GRCh38
rs149730601 Chr.17: 44148880-44148880 on Build GRCh38
rs745748184 Chr.17: 44148882-44148882 on Build GRCh38
rs766866793 Chr.17: 44148830-44148830 on Build GRCh38
rs774881553 Chr.17: 44148824-44148824 on Build GRCh38
rs779581894 Chr.17: 44148897-44148897 on Build GRCh38
rs916000044 Chr.17: 44148890-44148890 on Build GRCh38

View Full Product Details