Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57382236-57382489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CL1
Entrez Gene ID
6376
Gene Name
C-X3-C motif chemokine ligand 1
Gene Aliases
ABCD-3, C3Xkine, CXC3, CXC3C, NTN, NTT, SCYD1, fractalkine, neurotactin
UniGene
Hs.531668
Gene Chromosome Location
Chr.16: 57372461-57385048 on Build GRCh38
Transcripts
NM_001304392.1 NM_002996.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4755962 c.406A>G p.S136G Chr.16: 57382244-57382244 on Build GRCh38
2836359 c.647C>T p.S216F Chr.16: 57382485-57382485 on Build GRCh38
5628905 c.483G>A p.V161V Chr.16: 57382321-57382321 on Build GRCh38
5740659 c.435G>T p.Q145H Chr.16: 57382273-57382273 on Build GRCh38
3729650 c.593A>G p.Q198R Chr.16: 57382431-57382431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192202449 Chr.16: 57382408-57382408 on Build GRCh38
rs780537893 Chr.16: 57382375-57382375 on Build GRCh38
rs368032945 Chr.16: 57382315-57382315 on Build GRCh38
rs759450654 Chr.16: 57382409-57382409 on Build GRCh38
rs541210508 Chr.16: 57382350-57382350 on Build GRCh38
rs756707032 Chr.16: 57382447-57382447 on Build GRCh38
rs746549764 Chr.16: 57382393-57382393 on Build GRCh38
rs200354035 Chr.16: 57382257-57382257 on Build GRCh38
rs142113631 Chr.16: 57382414-57382414 on Build GRCh38
rs199869986 Chr.16: 57382353-57382353 on Build GRCh38
rs758713105 Chr.16: 57382339-57382339 on Build GRCh38
rs781755423 Chr.16: 57382259-57382259 on Build GRCh38
rs149956288 Chr.16: 57382349-57382349 on Build GRCh38
rs772762556 Chr.16: 57382312-57382312 on Build GRCh38
rs755255029 Chr.16: 57382461-57382461 on Build GRCh38
rs149081012 Chr.16: 57382418-57382418 on Build GRCh38
rs759347436 Chr.16: 57382318-57382318 on Build GRCh38
rs781770721 Chr.16: 57382351-57382351 on Build GRCh38
rs748916584 Chr.16: 57382416-57382416 on Build GRCh38
rs762000058 Chr.16: 57382426-57382426 on Build GRCh38
rs772817365 Chr.16: 57382387-57382387 on Build GRCh38
rs900186919 Chr.16: 57382402-57382402 on Build GRCh38
rs760152855 Chr.16: 57382314-57382314 on Build GRCh38
rs762624738 Chr.16: 57382417-57382417 on Build GRCh38
rs756443334 Chr.16: 57382369-57382369 on Build GRCh38
rs747773127 Chr.16: 57382309-57382309 on Build GRCh38
rs746604824 Chr.16: 57382464-57382464 on Build GRCh38
rs143145386 Chr.16: 57382462-57382462 on Build GRCh38
rs749606953 Chr.16: 57382383-57382383 on Build GRCh38
rs563993035 Chr.16: 57382421-57382421 on Build GRCh38
rs370435863 Chr.16: 57382258-57382258 on Build GRCh38
rs138964688 Chr.16: 57382319-57382319 on Build GRCh38
rs752865713 Chr.16: 57382338-57382338 on Build GRCh38
rs141389026 Chr.16: 57382300-57382300 on Build GRCh38
rs771623366 Chr.16: 57382311-57382311 on Build GRCh38
rs750530311 Chr.16: 57382444-57382444 on Build GRCh38
rs770148243 Chr.16: 57382279-57382279 on Build GRCh38
rs773891382 Chr.16: 57382306-57382306 on Build GRCh38
rs970008642 Chr.16: 57382269-57382269 on Build GRCh38
rs200427867 Chr.16: 57382415-57382415 on Build GRCh38
rs374854139 Chr.16: 57382377-57382377 on Build GRCh38
rs774384575 Chr.16: 57382396-57382402 on Build GRCh38
rs771801757 Chr.16: 57382386-57382386 on Build GRCh38
rs1004864770 Chr.16: 57382420-57382420 on Build GRCh38
rs764200214 Chr.16: 57382342-57382342 on Build GRCh38
rs754266085 Chr.16: 57382457-57382457 on Build GRCh38
rs746266707 Chr.16: 57382264-57382264 on Build GRCh38
rs780398414 Chr.16: 57382456-57382456 on Build GRCh38
rs751730918 Chr.16: 57382344-57382344 on Build GRCh38
rs368270552 Chr.16: 57382433-57382433 on Build GRCh38
rs766949258 Chr.16: 57382398-57382398 on Build GRCh38

View Full Product Details