Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 845587-845818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM110A
Entrez Gene ID
83541
Gene Name
family with sequence similarity 110 member A
Gene Aliases
C20orf55, F10, bA371L19.3
UniGene
Hs.574822
Gene Chromosome Location
Chr.20: 833697-846279 on Build GRCh38
Transcripts
NM_001042353.2 NM_001289145.1 NM_001289146.1 NM_001289147.1 NM_031424.5 NM_207121.4 XM_011529381.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5494037 c.817C>T p.R273C Chr.20: 845621-845621 on Build GRCh38
1413288 c.801C>T p.G267G Chr.20: 845605-845605 on Build GRCh38
6988835 c.856C>T p.Q286* Chr.20: 845660-845660 on Build GRCh38
1681483 c.803T>C p.V268A Chr.20: 845607-845607 on Build GRCh38
1029305 c.829G>A p.V277M Chr.20: 845633-845633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566352996 Chr.20: 845623-845623 on Build GRCh38
rs1019995642 Chr.20: 845738-845738 on Build GRCh38
rs754955051 Chr.20: 845706-845706 on Build GRCh38
rs770513776 Chr.20: 845730-845730 on Build GRCh38
rs77193875 Chr.20: 845778-845778 on Build GRCh38
rs760091575 Chr.20: 845742-845742 on Build GRCh38
rs768145575 Chr.20: 845741-845741 on Build GRCh38
rs760001857 Chr.20: 845631-845631 on Build GRCh38
rs765636905 Chr.20: 845656-845656 on Build GRCh38
rs758683222 Chr.20: 845666-845666 on Build GRCh38
rs372778762 Chr.20: 845694-845694 on Build GRCh38
rs920559350 Chr.20: 845791-845791 on Build GRCh38
rs772603265 Chr.20: 845606-845606 on Build GRCh38
rs6055290 Chr.20: 845654-845654 on Build GRCh38
rs375915338 Chr.20: 845697-845697 on Build GRCh38
rs150839256 Chr.20: 845713-845713 on Build GRCh38
rs762408732 Chr.20: 845708-845709 on Build GRCh38
rs746529741 Chr.20: 845723-845723 on Build GRCh38
rs772872188 Chr.20: 845715-845715 on Build GRCh38
rs943609985 Chr.20: 845781-845781 on Build GRCh38
rs775979407 Chr.20: 845737-845737 on Build GRCh38
rs955164752 Chr.20: 845740-845740 on Build GRCh38
rs535040807 Chr.20: 845700-845700 on Build GRCh38
rs753042458 Chr.20: 845664-845664 on Build GRCh38
rs975412443 Chr.20: 845783-845783 on Build GRCh38
rs762302254 Chr.20: 845641-845641 on Build GRCh38
rs201672012 Chr.20: 845714-845714 on Build GRCh38
rs760173723 Chr.20: 845612-845612 on Build GRCh38
rs868662309 Chr.20: 845622-845622 on Build GRCh38
rs751618067 Chr.20: 845674-845674 on Build GRCh38
rs759072121 Chr.20: 845773-845773 on Build GRCh38
rs757323332 Chr.20: 845689-845689 on Build GRCh38
rs766815462 Chr.20: 845618-845618 on Build GRCh38
rs764323987 Chr.20: 845667-845667 on Build GRCh38
rs556992798 Chr.20: 845792-845792 on Build GRCh38

View Full Product Details