Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 67900931-67901204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A1
Entrez Gene ID
84210
Gene Name
ankyrin repeat domain 20 family member A1
Gene Aliases
ANKRD20A
UniGene
Hs.632671
Gene Chromosome Location
Chr.9: 67859315-67902394 on Build GRCh38
Transcripts
NM_032250.3 XM_005272532.4

SNP Detail

SNP ID Chromosome Location
rs775141469 Chr.9: 67900983-67900983 on Build GRCh38
rs1008868367 Chr.9: 67901146-67901146 on Build GRCh38
rs200294490 Chr.9: 67901094-67901094 on Build GRCh38
rs768772389 Chr.9: 67900991-67900991 on Build GRCh38
rs202221455 Chr.9: 67901048-67901048 on Build GRCh38
rs751696814 Chr.9: 67901021-67901021 on Build GRCh38
rs62542472 Chr.9: 67901030-67901030 on Build GRCh38
rs774223199 Chr.9: 67900973-67900974 on Build GRCh38
rs748514293 Chr.9: 67900968-67900968 on Build GRCh38
rs775283222 Chr.9: 67900998-67900999 on Build GRCh38
rs745364158 Chr.9: 67900978-67900978 on Build GRCh38
rs201269458 Chr.9: 67901131-67901131 on Build GRCh38
rs745441337 Chr.9: 67901037-67901037 on Build GRCh38
rs779697965 Chr.9: 67901039-67901039 on Build GRCh38
rs770589545 Chr.9: 67900971-67900971 on Build GRCh38
rs756388246 Chr.9: 67901036-67901036 on Build GRCh38
rs771433566 Chr.9: 67900973-67900976 on Build GRCh38
rs776464488 Chr.9: 67900994-67900994 on Build GRCh38
rs765110091 Chr.9: 67901010-67901010 on Build GRCh38
rs771647050 Chr.9: 67900980-67900980 on Build GRCh38
rs761825410 Chr.9: 67901046-67901046 on Build GRCh38
rs769393213 Chr.9: 67900994-67900995 on Build GRCh38
rs761552057 Chr.9: 67900997-67900997 on Build GRCh38
rs778391734 Chr.9: 67900973-67900973 on Build GRCh38
rs577870702 Chr.9: 67901020-67901020 on Build GRCh38
rs762633335 Chr.9: 67900998-67901002 on Build GRCh38
rs746565167 Chr.9: 67901040-67901040 on Build GRCh38
rs763168362 Chr.9: 67901013-67901013 on Build GRCh38
rs745553890 Chr.9: 67900974-67900974 on Build GRCh38
rs538856468 Chr.9: 67901047-67901047 on Build GRCh38
rs1040791900 Chr.9: 67901174-67901174 on Build GRCh38
rs753281236 Chr.9: 67901035-67901035 on Build GRCh38
rs755460701 Chr.9: 67901022-67901022 on Build GRCh38
rs760681311 Chr.9: 67900988-67900988 on Build GRCh38
rs781527179 Chr.9: 67900964-67900964 on Build GRCh38
rs200972432 Chr.9: 67901012-67901012 on Build GRCh38
rs747494284 Chr.9: 67900964-67900964 on Build GRCh38

View Full Product Details