Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31655275-31655548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
UniGene
Hs.368921
Gene Chromosome Location
Chr.1: 31652247-31704167 on Build GRCh38
Transcripts
NM_001856.3 XM_005270481.1 XM_011540722.1 XM_011540723.1 XM_011540724.1 XM_011540726.2 XM_011540727.2 XM_011540728.2 XM_017000338.1 XM_017000339.1 XM_017000340.1 XM_017000341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6294472 c.4241C>A p.A1414D Chr.1: 31655363-31655363 on Build GRCh38
2077235 c.4107T>C p.D1369D Chr.1: 31655497-31655497 on Build GRCh38
6292467 c.4286C>G p.S1429C Chr.1: 31655318-31655318 on Build GRCh38
2077234 c.4156G>A p.G1386S Chr.1: 31655448-31655448 on Build GRCh38
2077233 c.4244C>T p.P1415L Chr.1: 31655360-31655360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749754753 Chr.1: 31655430-31655430 on Build GRCh38
rs199971611 Chr.1: 31655395-31655395 on Build GRCh38
rs570978287 Chr.1: 31655341-31655341 on Build GRCh38
rs758141855 Chr.1: 31655307-31655307 on Build GRCh38
rs751389279 Chr.1: 31655508-31655508 on Build GRCh38
rs932786201 Chr.1: 31655439-31655439 on Build GRCh38
rs774578187 Chr.1: 31655357-31655357 on Build GRCh38
rs551467305 Chr.1: 31655301-31655301 on Build GRCh38
rs534878932 Chr.1: 31655446-31655446 on Build GRCh38
rs771239207 Chr.1: 31655433-31655433 on Build GRCh38
rs368836139 Chr.1: 31655380-31655380 on Build GRCh38
rs779582351 Chr.1: 31655316-31655316 on Build GRCh38
rs755956023 Chr.1: 31655435-31655435 on Build GRCh38
rs374704588 Chr.1: 31655371-31655371 on Build GRCh38
rs201772427 Chr.1: 31655343-31655343 on Build GRCh38
rs746144920 Chr.1: 31655449-31655449 on Build GRCh38
rs752252028 Chr.1: 31655420-31655420 on Build GRCh38
rs1040746200 Chr.1: 31655412-31655412 on Build GRCh38
rs563458298 Chr.1: 31655387-31655387 on Build GRCh38
rs943729707 Chr.1: 31655383-31655383 on Build GRCh38
rs762920912 Chr.1: 31655488-31655488 on Build GRCh38
rs779469775 Chr.1: 31655360-31655360 on Build GRCh38
rs1048533847 Chr.1: 31655448-31655448 on Build GRCh38
rs200894307 Chr.1: 31655423-31655423 on Build GRCh38
rs374803719 Chr.1: 31655350-31655350 on Build GRCh38
rs772276743 Chr.1: 31655450-31655450 on Build GRCh38
rs766330027 Chr.1: 31655492-31655492 on Build GRCh38
rs368047384 Chr.1: 31655351-31655351 on Build GRCh38
rs1016010144 Chr.1: 31655319-31655319 on Build GRCh38
rs746607673 Chr.1: 31655321-31655321 on Build GRCh38
rs960606757 Chr.1: 31655410-31655410 on Build GRCh38
rs1023408400 Chr.1: 31655434-31655434 on Build GRCh38
rs145204978 Chr.1: 31655512-31655512 on Build GRCh38
rs372291805 Chr.1: 31655381-31655381 on Build GRCh38
rs184582359 Chr.1: 31655461-31655461 on Build GRCh38
rs754571533 Chr.1: 31655330-31655330 on Build GRCh38
rs773050682 Chr.1: 31655474-31655474 on Build GRCh38
rs570053297 Chr.1: 31655378-31655378 on Build GRCh38
rs34811700 Chr.1: 31655359-31655359 on Build GRCh38
rs746169503 Chr.1: 31655509-31655512 on Build GRCh38
rs768728564 Chr.1: 31655470-31655470 on Build GRCh38
rs377253155 Chr.1: 31655514-31655514 on Build GRCh38
rs202040813 Chr.1: 31655499-31655499 on Build GRCh38
rs765136510 Chr.1: 31655296-31655296 on Build GRCh38
rs375279584 Chr.1: 31655396-31655396 on Build GRCh38
rs775620665 Chr.1: 31655458-31655458 on Build GRCh38
rs192646338 Chr.1: 31655347-31655347 on Build GRCh38
rs770190648 Chr.1: 31655344-31655344 on Build GRCh38
rs369233724 Chr.1: 31655518-31655518 on Build GRCh38
rs199551630 Chr.1: 31655425-31655425 on Build GRCh38
rs758284097 Chr.1: 31655347-31655355 on Build GRCh38

View Full Product Details