Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6450272-6450505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD27
Entrez Gene ID
939
Gene Name
CD27 molecule
Gene Aliases
S152, S152. LPFS2, T14, TNFRSF7, Tp55
UniGene
Hs.355307
Gene Chromosome Location
Chr.12: 6444867-6451718 on Build GRCh38
Transcripts
NM_001242.4 XM_011521042.2 XM_017020232.1 XM_017020233.1 XM_017020234.1 NR_015382.2

Gene Symbol
CD27-AS1
Entrez Gene ID
678655
Gene Name
CD27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655224
Gene Chromosome Location
Chr.12: 6439001-6451517 on Build GRCh38
Transcripts
NM_001242.4 XM_011521042.2 XM_017020232.1 XM_017020233.1 XM_017020234.1 NR_015382.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4670435 c.387C>T p.T129T Chr.12: 6450291-6450291 on Build GRCh38
7153437 c.370C>T p.P124S Chr.12: 6450274-6450274 on Build GRCh38
5051870 c.392G>A p.R131Q Chr.12: 6450296-6450296 on Build GRCh38
6223014 c.381G>A p.S127S Chr.12: 6450285-6450285 on Build GRCh38
942550 c.394T>C p.S132P Chr.12: 6450298-6450298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144098443 Chr.12: 6450321-6450321 on Build GRCh38
rs556282612 Chr.12: 6450478-6450478 on Build GRCh38
rs764320210 Chr.12: 6450393-6450393 on Build GRCh38
rs776882476 Chr.12: 6450394-6450394 on Build GRCh38
rs906923884 Chr.12: 6450400-6450400 on Build GRCh38
rs539863789 Chr.12: 6450425-6450425 on Build GRCh38
rs745521506 Chr.12: 6450383-6450383 on Build GRCh38
rs887499800 Chr.12: 6450438-6450438 on Build GRCh38
rs143035415 Chr.12: 6450292-6450292 on Build GRCh38
rs756058870 Chr.12: 6450329-6450329 on Build GRCh38
rs377343532 Chr.12: 6450293-6450293 on Build GRCh38
rs373368586 Chr.12: 6450317-6450317 on Build GRCh38
rs763226101 Chr.12: 6450314-6450314 on Build GRCh38
rs771715961 Chr.12: 6450387-6450387 on Build GRCh38
rs765725985 Chr.12: 6450299-6450299 on Build GRCh38
rs747930868 Chr.12: 6450328-6450328 on Build GRCh38
rs771476874 Chr.12: 6450357-6450388 on Build GRCh38
rs897526946 Chr.12: 6450446-6450446 on Build GRCh38
rs773702298 Chr.12: 6450403-6450403 on Build GRCh38
rs765702056 Chr.12: 6450401-6450401 on Build GRCh38
rs975522364 Chr.12: 6450303-6450303 on Build GRCh38
rs766652363 Chr.12: 6450309-6450309 on Build GRCh38
rs199876113 Chr.12: 6450291-6450291 on Build GRCh38
rs748876729 Chr.12: 6450348-6450348 on Build GRCh38
rs898370779 Chr.12: 6450459-6450459 on Build GRCh38
rs941781474 Chr.12: 6450442-6450442 on Build GRCh38
rs750549892 Chr.12: 6450300-6450300 on Build GRCh38
rs150578547 Chr.12: 6450295-6450295 on Build GRCh38
rs564047552 Chr.12: 6450346-6450346 on Build GRCh38
rs762215347 Chr.12: 6450396-6450396 on Build GRCh38
rs1036014577 Chr.12: 6450444-6450444 on Build GRCh38
rs776987985 Chr.12: 6450296-6450296 on Build GRCh38
rs188225311 Chr.12: 6450364-6450364 on Build GRCh38
rs773943561 Chr.12: 6450380-6450380 on Build GRCh38
rs758440909 Chr.12: 6450302-6450302 on Build GRCh38
rs997430193 Chr.12: 6450453-6450453 on Build GRCh38

View Full Product Details