Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75495534-75495759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1
Entrez Gene ID
11010
Gene Name
GLI pathogenesis related 1
Gene Aliases
CRISP7, GLIPR, RTVP1
UniGene
Hs.205558
Gene Chromosome Location
Chr.12: 75480733-75501936 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7181902 c.563G>C p.R188T Chr.12: 75495606-75495606 on Build GRCh38
4412989 c.535G>A p.G179R Chr.12: 75495578-75495578 on Build GRCh38
4211285 c.534-1G>C p.? Chr.12: 75495576-75495576 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs923968612 Chr.12: 75495617-75495617 on Build GRCh38
rs374976311 Chr.12: 75495708-75495708 on Build GRCh38
rs769877918 Chr.12: 75495576-75495576 on Build GRCh38
rs769755274 Chr.12: 75495698-75495698 on Build GRCh38
rs562658998 Chr.12: 75495610-75495610 on Build GRCh38
rs761963810 Chr.12: 75495643-75495643 on Build GRCh38
rs199814144 Chr.12: 75495563-75495563 on Build GRCh38
rs144379806 Chr.12: 75495590-75495590 on Build GRCh38
rs754340327 Chr.12: 75495659-75495659 on Build GRCh38
rs752963427 Chr.12: 75495571-75495582 on Build GRCh38
rs138249204 Chr.12: 75495725-75495725 on Build GRCh38
rs746037511 Chr.12: 75495681-75495681 on Build GRCh38
rs372628259 Chr.12: 75495677-75495677 on Build GRCh38
rs558237723 Chr.12: 75495561-75495561 on Build GRCh38
rs767798989 Chr.12: 75495633-75495633 on Build GRCh38
rs764615725 Chr.12: 75495656-75495656 on Build GRCh38
rs202112160 Chr.12: 75495697-75495697 on Build GRCh38
rs150875077 Chr.12: 75495703-75495703 on Build GRCh38
rs1039322491 Chr.12: 75495680-75495680 on Build GRCh38
rs61545347 Chr.12: 75495572-75495572 on Build GRCh38
rs934046941 Chr.12: 75495618-75495618 on Build GRCh38
rs7137996 Chr.12: 75495562-75495562 on Build GRCh38
rs762497177 Chr.12: 75495601-75495601 on Build GRCh38
rs758708089 Chr.12: 75495692-75495692 on Build GRCh38
rs535573464 Chr.12: 75495720-75495721 on Build GRCh38
rs759249847 Chr.12: 75495624-75495624 on Build GRCh38
rs756938561 Chr.12: 75495573-75495574 on Build GRCh38
rs778927301 Chr.12: 75495679-75495679 on Build GRCh38
rs143959784 Chr.12: 75495648-75495648 on Build GRCh38
rs750071992 Chr.12: 75495715-75495716 on Build GRCh38
rs749244340 Chr.12: 75495704-75495704 on Build GRCh38
rs866515277 Chr.12: 75495595-75495595 on Build GRCh38
rs917065860 Chr.12: 75495598-75495598 on Build GRCh38
rs765859318 Chr.12: 75495609-75495609 on Build GRCh38
rs899100500 Chr.12: 75495688-75495688 on Build GRCh38
rs774435352 Chr.12: 75495596-75495596 on Build GRCh38
rs778478951 Chr.12: 75495632-75495634 on Build GRCh38

View Full Product Details