Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28534338-28534581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXN1
Entrez Gene ID
8456
Gene Name
forkhead box N1
Gene Aliases
FKHL20, RONU, WHN
UniGene
Hs.58611
Gene Chromosome Location
Chr.17: 28506211-28538900 on Build GRCh38
Transcripts
NM_003593.2 XM_005258046.3 XM_011525358.1 XM_011525359.1 XM_011525362.1 XM_011525367.1 XM_011525368.2 XM_011525369.2 XM_011525370.2 XM_017025228.1 XM_017025229.1 XM_017025230.1 XM_017025231.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880964 c.1135+1G>A p.? Chr.17: 28534539-28534539 on Build GRCh38
2800264 c.987C>T p.F329F Chr.17: 28534390-28534390 on Build GRCh38
436234 c.937G>A p.D313N Chr.17: 28534340-28534340 on Build GRCh38
2800265 c.1027G>A p.G343S Chr.17: 28534430-28534430 on Build GRCh38
3515385 c.1016C>T p.S339F Chr.17: 28534419-28534419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771407322 Chr.17: 28534452-28534452 on Build GRCh38
rs762513012 Chr.17: 28534462-28534462 on Build GRCh38
rs774123218 Chr.17: 28534481-28534481 on Build GRCh38
rs202021251 Chr.17: 28534395-28534395 on Build GRCh38
rs917639702 Chr.17: 28534522-28534522 on Build GRCh38
rs893690865 Chr.17: 28534525-28534525 on Build GRCh38
rs200567079 Chr.17: 28534557-28534557 on Build GRCh38
rs139218809 Chr.17: 28534534-28534534 on Build GRCh38
rs766273178 Chr.17: 28534519-28534519 on Build GRCh38
rs150671729 Chr.17: 28534453-28534453 on Build GRCh38
rs943085047 Chr.17: 28534479-28534479 on Build GRCh38
rs141965955 Chr.17: 28534424-28534424 on Build GRCh38
rs377224791 Chr.17: 28534558-28534558 on Build GRCh38
rs779985336 Chr.17: 28534378-28534378 on Build GRCh38
rs755005215 Chr.17: 28534541-28534541 on Build GRCh38
rs866929627 Chr.17: 28534454-28534454 on Build GRCh38
rs979365967 Chr.17: 28534456-28534456 on Build GRCh38
rs996815327 Chr.17: 28534413-28534413 on Build GRCh38
rs184956155 Chr.17: 28534390-28534390 on Build GRCh38
rs756849720 Chr.17: 28534425-28534425 on Build GRCh38
rs778010436 Chr.17: 28534548-28534548 on Build GRCh38
rs762643472 Chr.17: 28534422-28534422 on Build GRCh38
rs376778792 Chr.17: 28534442-28534442 on Build GRCh38
rs148161520 Chr.17: 28534400-28534400 on Build GRCh38
rs199983506 Chr.17: 28534391-28534391 on Build GRCh38
rs868232906 Chr.17: 28534375-28534375 on Build GRCh38
rs1031281442 Chr.17: 28534463-28534463 on Build GRCh38
rs761738547 Chr.17: 28534498-28534498 on Build GRCh38
rs867638414 Chr.17: 28534509-28534509 on Build GRCh38
rs747328174 Chr.17: 28534550-28534550 on Build GRCh38
rs756285596 Chr.17: 28534372-28534372 on Build GRCh38
rs771369671 Chr.17: 28534554-28534554 on Build GRCh38
rs767213983 Chr.17: 28534510-28534510 on Build GRCh38
rs779158182 Chr.17: 28534543-28534543 on Build GRCh38
rs369196717 Chr.17: 28534547-28534547 on Build GRCh38
rs933053205 Chr.17: 28534396-28534396 on Build GRCh38
rs2286521 Chr.17: 28534546-28534546 on Build GRCh38
rs763859965 Chr.17: 28534467-28534467 on Build GRCh38
rs777145444 Chr.17: 28534556-28534556 on Build GRCh38
rs371817664 Chr.17: 28534518-28534518 on Build GRCh38
rs750434623 Chr.17: 28534517-28534517 on Build GRCh38

View Full Product Details