Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17213622-17213893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLCN
Entrez Gene ID
201163
Gene Name
folliculin
Gene Aliases
BHD, FLCL
UniGene
Hs.31652
Gene Chromosome Location
Chr.17: 17206945-17237188 on Build GRCh38
Transcripts
NM_144997.5 XM_011523714.2 XM_011523716.1 XM_011523718.2 XM_011523719.2 XM_011523721.2 XM_017024304.1 XM_017024305.1 XM_017024306.1 XM_017024307.1 XM_017024308.1 XM_017024309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163080 c.1663A>G p.T555A Chr.17: 17213732-17213732 on Build GRCh38
6871559 c.1608G>A p.L536L Chr.17: 17213787-17213787 on Build GRCh38
1381202 c.1622C>T p.A541V Chr.17: 17213773-17213773 on Build GRCh38
6580136 c.1677G>C p.K559N Chr.17: 17213718-17213718 on Build GRCh38
6978434 c.1587A>T p.K529N Chr.17: 17213808-17213808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748337450 Chr.17: 17213693-17213693 on Build GRCh38
rs878855215 Chr.17: 17213726-17213726 on Build GRCh38
rs775107483 Chr.17: 17213663-17213663 on Build GRCh38
rs777826268 Chr.17: 17213815-17213815 on Build GRCh38
rs980319370 Chr.17: 17213827-17213827 on Build GRCh38
rs201056799 Chr.17: 17213686-17213686 on Build GRCh38
rs398124531 Chr.17: 17213868-17213872 on Build GRCh38
rs201810397 Chr.17: 17213703-17213703 on Build GRCh38
rs774091922 Chr.17: 17213652-17213652 on Build GRCh38
rs755548579 Chr.17: 17213775-17213775 on Build GRCh38
rs781733528 Chr.17: 17213692-17213692 on Build GRCh38
rs773168989 Chr.17: 17213647-17213647 on Build GRCh38
rs879255684 Chr.17: 17213737-17213737 on Build GRCh38
rs767479616 Chr.17: 17213727-17213728 on Build GRCh38
rs753313171 Chr.17: 17213677-17213677 on Build GRCh38
rs760307957 Chr.17: 17213670-17213670 on Build GRCh38
rs749359334 Chr.17: 17213710-17213710 on Build GRCh38
rs876660810 Chr.17: 17213797-17213798 on Build GRCh38
rs774247151 Chr.17: 17213728-17213728 on Build GRCh38
rs978355107 Chr.17: 17213748-17213748 on Build GRCh38
rs767345167 Chr.17: 17213658-17213658 on Build GRCh38
rs756318617 Chr.17: 17213805-17213805 on Build GRCh38
rs368472222 Chr.17: 17213867-17213867 on Build GRCh38
rs771847652 Chr.17: 17213751-17213751 on Build GRCh38
rs779028759 Chr.17: 17213862-17213862 on Build GRCh38
rs878855214 Chr.17: 17213858-17213858 on Build GRCh38
rs759292330 Chr.17: 17213654-17213654 on Build GRCh38
rs760329266 Chr.17: 17213760-17213760 on Build GRCh38
rs745859406 Chr.17: 17213742-17213742 on Build GRCh38
rs886041478 Chr.17: 17213855-17213855 on Build GRCh38
rs771038343 Chr.17: 17213718-17213718 on Build GRCh38
rs879255683 Chr.17: 17213816-17213816 on Build GRCh38
rs876658649 Chr.17: 17213861-17213861 on Build GRCh38
rs147554296 Chr.17: 17213691-17213691 on Build GRCh38
rs398124532 Chr.17: 17213798-17213798 on Build GRCh38
rs758901704 Chr.17: 17213678-17213678 on Build GRCh38
rs776389684 Chr.17: 17213768-17213768 on Build GRCh38
rs1048214486 Chr.17: 17213662-17213662 on Build GRCh38
rs752161850 Chr.17: 17213687-17213687 on Build GRCh38
rs763688092 Chr.17: 17213763-17213763 on Build GRCh38
rs770795599 Chr.17: 17213651-17213651 on Build GRCh38
rs756302545 Chr.17: 17213698-17213698 on Build GRCh38
rs185419942 Chr.17: 17213772-17213772 on Build GRCh38
rs886039371 Chr.17: 17213794-17213794 on Build GRCh38
rs752108683 Chr.17: 17213774-17213774 on Build GRCh38
rs764899882 Chr.17: 17213773-17213773 on Build GRCh38
rs753023144 Chr.17: 17213801-17213801 on Build GRCh38
rs142288285 Chr.17: 17213836-17213836 on Build GRCh38
rs886426221 Chr.17: 17213666-17213666 on Build GRCh38
rs1057518043 Chr.17: 17213796-17213817 on Build GRCh38
rs1022566650 Chr.17: 17213697-17213697 on Build GRCh38
rs775149348 Chr.17: 17213758-17213758 on Build GRCh38
rs757312148 Chr.17: 17213859-17213859 on Build GRCh38
rs753009073 Chr.17: 17213815-17213816 on Build GRCh38
rs763604691 Chr.17: 17213671-17213671 on Build GRCh38
rs190965235 Chr.17: 17213796-17213796 on Build GRCh38

View Full Product Details