Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 890198-890443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
UniGene
Hs.278973
Gene Chromosome Location
Chr.20: 869899-916367 on Build GRCh38
Transcripts
NM_001322809.1 NM_015985.3 XM_011529239.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6094005 c.457G>T p.E153* Chr.20: 890221-890221 on Build GRCh38
4748370 c.449C>A p.T150N Chr.20: 890229-890229 on Build GRCh38
3841591 c.336G>C p.L112F Chr.20: 890342-890342 on Build GRCh38
385914 c.460G>T p.A154S Chr.20: 890218-890218 on Build GRCh38
3423840 c.341C>T p.S114L Chr.20: 890337-890337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143679853 Chr.20: 890294-890294 on Build GRCh38
rs202046919 Chr.20: 890257-890257 on Build GRCh38
rs749890952 Chr.20: 890419-890419 on Build GRCh38
rs746352961 Chr.20: 890290-890290 on Build GRCh38
rs927121483 Chr.20: 890266-890266 on Build GRCh38
rs370865591 Chr.20: 890345-890345 on Build GRCh38
rs748884201 Chr.20: 890407-890407 on Build GRCh38
rs564494099 Chr.20: 890291-890291 on Build GRCh38
rs13038240 Chr.20: 890369-890369 on Build GRCh38
rs780729817 Chr.20: 890358-890358 on Build GRCh38
rs150123876 Chr.20: 890238-890238 on Build GRCh38
rs746455441 Chr.20: 890281-890281 on Build GRCh38
rs79320190 Chr.20: 890327-890327 on Build GRCh38
rs761948894 Chr.20: 890316-890316 on Build GRCh38
rs766941962 Chr.20: 890415-890415 on Build GRCh38
rs868514078 Chr.20: 890222-890222 on Build GRCh38
rs775920001 Chr.20: 890292-890292 on Build GRCh38
rs375449785 Chr.20: 890396-890396 on Build GRCh38
rs753702124 Chr.20: 890251-890251 on Build GRCh38
rs200858753 Chr.20: 890346-890346 on Build GRCh38
rs370322294 Chr.20: 890243-890243 on Build GRCh38
rs770243314 Chr.20: 890286-890286 on Build GRCh38
rs1041399657 Chr.20: 890304-890304 on Build GRCh38
rs117281778 Chr.20: 890393-890393 on Build GRCh38
rs946781468 Chr.20: 890412-890412 on Build GRCh38
rs139958114 Chr.20: 890283-890283 on Build GRCh38
rs13038328 Chr.20: 890263-890263 on Build GRCh38
rs769034261 Chr.20: 890382-890382 on Build GRCh38
rs774505929 Chr.20: 890239-890239 on Build GRCh38
rs758039784 Chr.20: 890276-890276 on Build GRCh38
rs201472997 Chr.20: 890395-890395 on Build GRCh38
rs140621916 Chr.20: 890348-890348 on Build GRCh38
rs1047505618 Chr.20: 890297-890297 on Build GRCh38
rs762695083 Chr.20: 890403-890403 on Build GRCh38
rs763225404 Chr.20: 890228-890228 on Build GRCh38
rs759501738 Chr.20: 890331-890331 on Build GRCh38
rs981949928 Chr.20: 890425-890425 on Build GRCh38
rs541174884 Chr.20: 890272-890272 on Build GRCh38
rs376414024 Chr.20: 890337-890337 on Build GRCh38
rs760702359 Chr.20: 890240-890240 on Build GRCh38
rs749527768 Chr.20: 890293-890293 on Build GRCh38
rs751261151 Chr.20: 890407-890407 on Build GRCh38
rs201928850 Chr.20: 890379-890379 on Build GRCh38
rs779322497 Chr.20: 890387-890387 on Build GRCh38
rs376079007 Chr.20: 890227-890227 on Build GRCh38
rs756579841 Chr.20: 890371-890371 on Build GRCh38
rs943789944 Chr.20: 890336-890336 on Build GRCh38
rs942929399 Chr.20: 890408-890408 on Build GRCh38
rs13037050 Chr.20: 890280-890280 on Build GRCh38
rs767678821 Chr.20: 890322-890322 on Build GRCh38
rs138675528 Chr.20: 890218-890218 on Build GRCh38
rs76996146 Chr.20: 890224-890224 on Build GRCh38
rs576213874 Chr.20: 890219-890219 on Build GRCh38
rs781511303 Chr.20: 890284-890284 on Build GRCh38
rs71191984 Chr.20: 890425-890426 on Build GRCh38
rs200194275 Chr.20: 890406-890406 on Build GRCh38
rs370007562 Chr.20: 890411-890411 on Build GRCh38
rs542780866 Chr.20: 890363-890363 on Build GRCh38
rs780295979 Chr.20: 890378-890378 on Build GRCh38
rs13043898 Chr.20: 890268-890268 on Build GRCh38
rs775169357 Chr.20: 890295-890295 on Build GRCh38
rs142196008 Chr.20: 890349-890349 on Build GRCh38
rs777266158 Chr.20: 890278-890278 on Build GRCh38

View Full Product Details