Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142457491-142457720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATR
Entrez Gene ID
545
Gene Name
ATR serine/threonine kinase
Gene Aliases
FCTCS, FRP1, MEC1, SCKL, SCKL1
UniGene
Hs.271791
Gene Chromosome Location
Chr.3: 142449235-142578826 on Build GRCh38
Transcripts
NM_001184.3 XM_011512924.1 XM_011512925.1 XM_017006643.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
137863 c.7646C>T p.P2549L Chr.3: 142457613-142457613 on Build GRCh38
6908797 c.7623G>T p.R2541S Chr.3: 142457636-142457636 on Build GRCh38
5777106 c.7632T>C p.R2544R Chr.3: 142457627-142457627 on Build GRCh38
6587889 c.7545G>C p.L2515L Chr.3: 142457714-142457714 on Build GRCh38
4114118 c.7640G>A p.R2547Q Chr.3: 142457619-142457619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951775769 Chr.3: 142457541-142457541 on Build GRCh38
rs768497377 Chr.3: 142457608-142457608 on Build GRCh38
rs914247885 Chr.3: 142457671-142457671 on Build GRCh38
rs765583100 Chr.3: 142457642-142457642 on Build GRCh38
rs762108631 Chr.3: 142457638-142457638 on Build GRCh38
rs745414333 Chr.3: 142457563-142457563 on Build GRCh38
rs769121210 Chr.3: 142457628-142457628 on Build GRCh38
rs915275959 Chr.3: 142457639-142457639 on Build GRCh38
rs763127779 Chr.3: 142457661-142457661 on Build GRCh38
rs747465705 Chr.3: 142457569-142457569 on Build GRCh38
rs542358200 Chr.3: 142457528-142457528 on Build GRCh38
rs746979167 Chr.3: 142457587-142457587 on Build GRCh38
rs556927325 Chr.3: 142457543-142457543 on Build GRCh38
rs773381675 Chr.3: 142457654-142457654 on Build GRCh38
rs369795894 Chr.3: 142457570-142457570 on Build GRCh38
rs983226197 Chr.3: 142457550-142457550 on Build GRCh38
rs908690726 Chr.3: 142457552-142457552 on Build GRCh38
rs554620139 Chr.3: 142457549-142457549 on Build GRCh38
rs771838121 Chr.3: 142457564-142457564 on Build GRCh38
rs558955623 Chr.3: 142457593-142457593 on Build GRCh38
rs976274149 Chr.3: 142457548-142457548 on Build GRCh38
rs144427735 Chr.3: 142457627-142457627 on Build GRCh38

View Full Product Details