Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102878780-102879053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AS3MT
Entrez Gene ID
57412
Gene Name
arsenite methyltransferase
Gene Aliases
CYT19
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102869453-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Gene Symbol
BORCS7-ASMT
Entrez Gene ID
100528007
Gene Name
BORCS7-ASMT readthrough (NMD candidate)
Gene Aliases
C10orf32-AS3MT, C10orf32-ASMT
UniGene
Hs.720370
Gene Chromosome Location
Chr.10: 102854210-102901899 on Build GRCh38
Transcripts
NM_020682.3 NR_037644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1345518 c.858A>C p.L286L Chr.10: 102878964-102878964 on Build GRCh38
6052693 c.752G>A p.R251H Chr.10: 102878858-102878858 on Build GRCh38
6064772 c.773G>C p.R258P Chr.10: 102878879-102878879 on Build GRCh38
6128134 c.743G>T p.G248V Chr.10: 102878849-102878849 on Build GRCh38
3709816 c.838A>G p.T280A Chr.10: 102878944-102878944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758809334 Chr.10: 102878991-102878991 on Build GRCh38
rs756083178 Chr.10: 102878904-102878905 on Build GRCh38
rs897394438 Chr.10: 102878813-102878813 on Build GRCh38
rs372316717 Chr.10: 102878978-102878978 on Build GRCh38
rs748589961 Chr.10: 102878857-102878857 on Build GRCh38
rs956599944 Chr.10: 102879026-102879026 on Build GRCh38
rs766279552 Chr.10: 102878908-102878909 on Build GRCh38
rs781323286 Chr.10: 102878894-102878895 on Build GRCh38
rs755252196 Chr.10: 102878975-102878975 on Build GRCh38
rs139656545 Chr.10: 102878858-102878858 on Build GRCh38
rs201755295 Chr.10: 102879034-102879034 on Build GRCh38
rs753453310 Chr.10: 102879031-102879031 on Build GRCh38
rs767199575 Chr.10: 102878802-102878802 on Build GRCh38
rs747427312 Chr.10: 102878893-102878893 on Build GRCh38
rs775241830 Chr.10: 102878901-102878901 on Build GRCh38
rs967902054 Chr.10: 102878986-102878986 on Build GRCh38
rs762696583 Chr.10: 102878902-102878902 on Build GRCh38
rs771442515 Chr.10: 102878894-102878894 on Build GRCh38
rs754984848 Chr.10: 102878839-102878839 on Build GRCh38
rs753400543 Chr.10: 102878912-102878913 on Build GRCh38
rs1000681263 Chr.10: 102879018-102879018 on Build GRCh38
rs377369423 Chr.10: 102878848-102878848 on Build GRCh38
rs11191439 Chr.10: 102878966-102878966 on Build GRCh38
rs781697893 Chr.10: 102879010-102879010 on Build GRCh38
rs779064702 Chr.10: 102878854-102878854 on Build GRCh38
rs768417929 Chr.10: 102879019-102879019 on Build GRCh38
rs766168360 Chr.10: 102878962-102878962 on Build GRCh38
rs750353278 Chr.10: 102878939-102878939 on Build GRCh38
rs778291892 Chr.10: 102878996-102878996 on Build GRCh38
rs760424988 Chr.10: 102878947-102878947 on Build GRCh38
rs1006659023 Chr.10: 102878967-102878967 on Build GRCh38
rs754093819 Chr.10: 102878828-102878828 on Build GRCh38
rs750275455 Chr.10: 102878804-102878804 on Build GRCh38
rs367745902 Chr.10: 102879023-102879023 on Build GRCh38
rs754476200 Chr.10: 102878994-102878998 on Build GRCh38
rs375414003 Chr.10: 102879004-102879004 on Build GRCh38
rs750357447 Chr.10: 102878842-102878842 on Build GRCh38
rs752861775 Chr.10: 102878989-102878989 on Build GRCh38
rs747548218 Chr.10: 102878997-102878997 on Build GRCh38
rs756311102 Chr.10: 102878818-102878818 on Build GRCh38
rs760440040 Chr.10: 102878827-102878827 on Build GRCh38
rs768221077 Chr.10: 102878909-102878909 on Build GRCh38
rs961628081 Chr.10: 102878806-102878806 on Build GRCh38
rs767544400 Chr.10: 102878934-102878934 on Build GRCh38
rs559312927 Chr.10: 102878889-102878889 on Build GRCh38
rs773965312 Chr.10: 102878912-102878912 on Build GRCh38
rs761629464 Chr.10: 102878926-102878926 on Build GRCh38
rs370398092 Chr.10: 102878801-102878801 on Build GRCh38
rs544746006 Chr.10: 102879011-102879011 on Build GRCh38
rs879655435 Chr.10: 102879032-102879032 on Build GRCh38

View Full Product Details