Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46787018-46787267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESD
Entrez Gene ID
2098
Gene Name
esterase D
Gene Aliases
FGH
UniGene
Hs.432491
Gene Chromosome Location
Chr.13: 46771256-46797681 on Build GRCh38
Transcripts
NM_001984.1 XM_005266278.2 XM_011534954.1 XM_017020414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1188712 c.98C>G p.A33G Chr.13: 46787080-46787080 on Build GRCh38
6021216 c.90G>A p.M30I Chr.13: 46787088-46787088 on Build GRCh38
79804 c.144G>T p.L48L Chr.13: 46787034-46787034 on Build GRCh38
6656305 c.135C>T p.C45C Chr.13: 46787043-46787043 on Build GRCh38
1942151 c.150G>A p.W50* Chr.13: 46787028-46787028 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs981063617 Chr.13: 46787164-46787164 on Build GRCh38
rs775233009 Chr.13: 46787124-46787126 on Build GRCh38
rs780388207 Chr.13: 46787080-46787080 on Build GRCh38
rs768882465 Chr.13: 46787111-46787111 on Build GRCh38
rs376968661 Chr.13: 46787148-46787148 on Build GRCh38
rs368378364 Chr.13: 46787112-46787112 on Build GRCh38
rs922365868 Chr.13: 46787173-46787176 on Build GRCh38
rs781400478 Chr.13: 46787056-46787056 on Build GRCh38
rs767348316 Chr.13: 46787067-46787068 on Build GRCh38
rs560713128 Chr.13: 46787191-46787191 on Build GRCh38
rs779398635 Chr.13: 46787142-46787142 on Build GRCh38
rs189058725 Chr.13: 46787204-46787204 on Build GRCh38
rs761209871 Chr.13: 46787206-46787206 on Build GRCh38
rs750445758 Chr.13: 46787069-46787069 on Build GRCh38
rs771788083 Chr.13: 46787149-46787149 on Build GRCh38
rs756212963 Chr.13: 46787079-46787079 on Build GRCh38
rs749844457 Chr.13: 46787086-46787086 on Build GRCh38
rs910058988 Chr.13: 46787042-46787042 on Build GRCh38

View Full Product Details