Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81254419-81254660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618356 c.443C>T p.S148L Chr.5: 81254487-81254487 on Build GRCh38
3618355 c.397G>A p.G133S Chr.5: 81254441-81254441 on Build GRCh38
1201256 c.376G>A p.V126I Chr.5: 81254420-81254420 on Build GRCh38
6629613 c.430G>A p.D144N Chr.5: 81254474-81254474 on Build GRCh38
5524748 c.429G>A p.T143T Chr.5: 81254473-81254473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566303812 Chr.5: 81254637-81254637 on Build GRCh38
rs759752699 Chr.5: 81254500-81254500 on Build GRCh38
rs375630161 Chr.5: 81254541-81254541 on Build GRCh38
rs908898708 Chr.5: 81254547-81254547 on Build GRCh38
rs988508366 Chr.5: 81254565-81254565 on Build GRCh38
rs1009786036 Chr.5: 81254591-81254591 on Build GRCh38
rs761016053 Chr.5: 81254537-81254537 on Build GRCh38
rs777664341 Chr.5: 81254449-81254449 on Build GRCh38
rs372834193 Chr.5: 81254522-81254522 on Build GRCh38
rs762372173 Chr.5: 81254526-81254526 on Build GRCh38
rs965661387 Chr.5: 81254574-81254574 on Build GRCh38
rs957629272 Chr.5: 81254561-81254561 on Build GRCh38
rs752790211 Chr.5: 81254565-81254565 on Build GRCh38
rs891343817 Chr.5: 81254590-81254590 on Build GRCh38
rs1020888464 Chr.5: 81254614-81254614 on Build GRCh38
rs769313745 Chr.5: 81254510-81254510 on Build GRCh38
rs34330118 Chr.5: 81254545-81254545 on Build GRCh38
rs146777847 Chr.5: 81254473-81254473 on Build GRCh38
rs773392416 Chr.5: 81254530-81254530 on Build GRCh38
rs759486668 Chr.5: 81254544-81254544 on Build GRCh38
rs767396239 Chr.5: 81254599-81254599 on Build GRCh38
rs758539278 Chr.5: 81254573-81254573 on Build GRCh38
rs752468928 Chr.5: 81254622-81254622 on Build GRCh38
rs899991147 Chr.5: 81254566-81254566 on Build GRCh38
rs139195835 Chr.5: 81254629-81254629 on Build GRCh38
rs757073013 Chr.5: 81254472-81254472 on Build GRCh38
rs529542764 Chr.5: 81254620-81254620 on Build GRCh38
rs112573022 Chr.5: 81254517-81254517 on Build GRCh38
rs563844338 Chr.5: 81254575-81254575 on Build GRCh38
rs968099026 Chr.5: 81254627-81254627 on Build GRCh38
rs751447217 Chr.5: 81254454-81254454 on Build GRCh38
rs199944303 Chr.5: 81254521-81254521 on Build GRCh38
rs941759664 Chr.5: 81254552-81254552 on Build GRCh38
rs779686334 Chr.5: 81254514-81254514 on Build GRCh38
rs772571263 Chr.5: 81254529-81254529 on Build GRCh38

View Full Product Details