Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47562575-47562846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG5
Entrez Gene ID
10675
Gene Name
chondroitin sulfate proteoglycan 5
Gene Aliases
NGC
UniGene
Hs.45127
Gene Chromosome Location
Chr.3: 47562238-47580240 on Build GRCh38
Transcripts
NM_001206942.1 NM_001206943.1 NM_001206944.1 NM_001206945.1 NM_006574.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3594786 c.1547C>T p.S516L Chr.3: 47562673-47562673 on Build GRCh38
4890315 c.1515G>T p.E505D Chr.3: 47562705-47562705 on Build GRCh38
5634767 c.1662T>G p.A554A Chr.3: 47562639-47562639 on Build GRCh38
3594788 c.1509G>A p.L503L Chr.3: 47562711-47562711 on Build GRCh38
4863798 c.1563A>C p.K521N Chr.3: 47562738-47562738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746049958 Chr.3: 47562789-47562790 on Build GRCh38
rs368217624 Chr.3: 47562781-47562782 on Build GRCh38
rs768667696 Chr.3: 47562662-47562662 on Build GRCh38
rs376512451 Chr.3: 47562790-47562791 on Build GRCh38
rs770435002 Chr.3: 47562673-47562673 on Build GRCh38
rs4858839 Chr.3: 47562809-47562809 on Build GRCh38
rs548312605 Chr.3: 47562786-47562786 on Build GRCh38
rs753385640 Chr.3: 47562817-47562817 on Build GRCh38
rs547163216 Chr.3: 47562781-47562781 on Build GRCh38
rs759252516 Chr.3: 47562728-47562728 on Build GRCh38
rs371021236 Chr.3: 47562672-47562672 on Build GRCh38
rs376844246 Chr.3: 47562783-47562783 on Build GRCh38
rs199986083 Chr.3: 47562789-47562789 on Build GRCh38
rs148021711 Chr.3: 47562679-47562679 on Build GRCh38
rs185202804 Chr.3: 47562787-47562787 on Build GRCh38
rs751580040 Chr.3: 47562770-47562771 on Build GRCh38
rs756074107 Chr.3: 47562615-47562615 on Build GRCh38
rs779011006 Chr.3: 47562784-47562785 on Build GRCh38
rs758112650 Chr.3: 47562806-47562806 on Build GRCh38
rs532154577 Chr.3: 47562700-47562700 on Build GRCh38
rs773841687 Chr.3: 47562688-47562688 on Build GRCh38
rs1023043517 Chr.3: 47562788-47562788 on Build GRCh38
rs9842590 Chr.3: 47562782-47562782 on Build GRCh38
rs772452394 Chr.3: 47562795-47562796 on Build GRCh38
rs534825971 Chr.3: 47562780-47562781 on Build GRCh38
rs753774150 Chr.3: 47562774-47562774 on Build GRCh38
rs116232180 Chr.3: 47562596-47562596 on Build GRCh38
rs777623634 Chr.3: 47562635-47562635 on Build GRCh38
rs377714163 Chr.3: 47562640-47562640 on Build GRCh38
rs994451752 Chr.3: 47562777-47562777 on Build GRCh38
rs779949815 Chr.3: 47562649-47562649 on Build GRCh38
rs751351353 Chr.3: 47562747-47562747 on Build GRCh38
rs1026774335 Chr.3: 47562684-47562684 on Build GRCh38
rs796421907 Chr.3: 47562781-47562782 on Build GRCh38
rs764081281 Chr.3: 47562766-47562766 on Build GRCh38
rs898945824 Chr.3: 47562764-47562764 on Build GRCh38
rs767034065 Chr.3: 47562729-47562729 on Build GRCh38
rs776797099 Chr.3: 47562671-47562671 on Build GRCh38
rs1047981703 Chr.3: 47562796-47562796 on Build GRCh38
rs144390955 Chr.3: 47562781-47562781 on Build GRCh38
rs369537090 Chr.3: 47562794-47562794 on Build GRCh38
rs766227760 Chr.3: 47562699-47562701 on Build GRCh38
rs371137351 Chr.3: 47562785-47562785 on Build GRCh38
rs748349383 Chr.3: 47562812-47562812 on Build GRCh38
rs564870179 Chr.3: 47562658-47562658 on Build GRCh38
rs138758003 Chr.3: 47562689-47562689 on Build GRCh38
rs755566544 Chr.3: 47562763-47562763 on Build GRCh38
rs758287540 Chr.3: 47562628-47562628 on Build GRCh38
rs765046308 Chr.3: 47562790-47562790 on Build GRCh38
rs773919844 Chr.3: 47562723-47562723 on Build GRCh38
rs773894819 Chr.3: 47562784-47562784 on Build GRCh38
rs770703185 Chr.3: 47562642-47562642 on Build GRCh38
rs752259042 Chr.3: 47562735-47562735 on Build GRCh38
rs969186915 Chr.3: 47562771-47562771 on Build GRCh38

View Full Product Details