Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16029620-16029893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKA
Entrez Gene ID
1187
Gene Name
chloride voltage-gated channel Ka
Gene Aliases
CLCK1, ClC-K1, hClC-Ka
UniGene
Hs.591533
Gene Chromosome Location
Chr.1: 16021991-16034050 on Build GRCh38
Transcripts
NM_001042704.1 NM_001257139.1 NM_004070.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6443755 c.1234A>T p.M412L Chr.1: 16029737-16029737 on Build GRCh38
6269875 c.1297+5G>T p.? Chr.1: 16029805-16029805 on Build GRCh38
5889479 c.1259C>T p.P420L Chr.1: 16029762-16029762 on Build GRCh38
1316869 c.1291A>G p.I431V Chr.1: 16029794-16029794 on Build GRCh38
1238525 c.1256T>C p.I419T Chr.1: 16029759-16029759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981296123 Chr.1: 16029869-16029869 on Build GRCh38
rs1010079764 Chr.1: 16029817-16029817 on Build GRCh38
rs1972359 Chr.1: 16029692-16029692 on Build GRCh38
rs371139669 Chr.1: 16029819-16029819 on Build GRCh38
rs780713062 Chr.1: 16029695-16029695 on Build GRCh38
rs548811846 Chr.1: 16029851-16029851 on Build GRCh38
rs771326944 Chr.1: 16029820-16029820 on Build GRCh38
rs778253967 Chr.1: 16029811-16029811 on Build GRCh38
rs542083067 Chr.1: 16029680-16029680 on Build GRCh38
rs756161038 Chr.1: 16029822-16029823 on Build GRCh38
rs200828311 Chr.1: 16029757-16029757 on Build GRCh38
rs1040312520 Chr.1: 16029737-16029737 on Build GRCh38
rs763082737 Chr.1: 16029774-16029774 on Build GRCh38
rs777833564 Chr.1: 16029691-16029691 on Build GRCh38
rs202220216 Chr.1: 16029763-16029763 on Build GRCh38
rs748763277 Chr.1: 16029711-16029711 on Build GRCh38
rs374555090 Chr.1: 16029835-16029835 on Build GRCh38
rs751859118 Chr.1: 16029774-16029776 on Build GRCh38
rs748818568 Chr.1: 16029803-16029803 on Build GRCh38
rs769319638 Chr.1: 16029708-16029708 on Build GRCh38
rs776728860 Chr.1: 16029728-16029728 on Build GRCh38
rs552108119 Chr.1: 16029802-16029802 on Build GRCh38
rs767704116 Chr.1: 16029803-16029803 on Build GRCh38
rs368912741 Chr.1: 16029720-16029720 on Build GRCh38
rs751477035 Chr.1: 16029782-16029782 on Build GRCh38
rs745469345 Chr.1: 16029703-16029703 on Build GRCh38
rs777118413 Chr.1: 16029824-16029824 on Build GRCh38
rs763130135 Chr.1: 16029845-16029845 on Build GRCh38
rs750475610 Chr.1: 16029809-16029809 on Build GRCh38
rs373111856 Chr.1: 16029805-16029805 on Build GRCh38
rs200979677 Chr.1: 16029833-16029833 on Build GRCh38
rs370236498 Chr.1: 16029773-16029773 on Build GRCh38
rs757017239 Chr.1: 16029783-16029783 on Build GRCh38
rs757410908 Chr.1: 16029799-16029800 on Build GRCh38
rs750181872 Chr.1: 16029797-16029797 on Build GRCh38
rs772615418 Chr.1: 16029713-16029713 on Build GRCh38
rs532394987 Chr.1: 16029772-16029772 on Build GRCh38
rs544165534 Chr.1: 16029709-16029709 on Build GRCh38
rs755830633 Chr.1: 16029798-16029798 on Build GRCh38
rs759947775 Chr.1: 16029844-16029844 on Build GRCh38
rs760900946 Chr.1: 16029674-16029674 on Build GRCh38
rs764162225 Chr.1: 16029775-16029775 on Build GRCh38
rs201976704 Chr.1: 16029758-16029758 on Build GRCh38
rs138144156 Chr.1: 16029693-16029693 on Build GRCh38
rs781101726 Chr.1: 16029794-16029794 on Build GRCh38
rs773659264 Chr.1: 16029718-16029718 on Build GRCh38
rs61772370 Chr.1: 16029657-16029657 on Build GRCh38

View Full Product Details