Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 160569281-160569526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10B
Entrez Gene ID
23120
Gene Name
ATPase phospholipid transporting 10B (putative)
Gene Aliases
ATPVB
UniGene
Hs.109358
Gene Chromosome Location
Chr.5: 160563120-160929411 on Build GRCh38
Transcripts
NM_025153.2 XM_006714833.2 XM_011534468.2 XM_011534469.1 XM_011534472.2 XM_017009252.1 XM_017009253.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4571157 c.3923T>C p.V1308A Chr.5: 160569511-160569511 on Build GRCh38
1065219 c.3910C>A p.L1304I Chr.5: 160569524-160569524 on Build GRCh38
5647148 c.3913A>G p.T1305A Chr.5: 160569521-160569521 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935645104 Chr.5: 160569431-160569431 on Build GRCh38
rs770102495 Chr.5: 160569451-160569451 on Build GRCh38
rs1036953199 Chr.5: 160569393-160569393 on Build GRCh38
rs368706700 Chr.5: 160569458-160569458 on Build GRCh38
rs775044803 Chr.5: 160569488-160569488 on Build GRCh38
rs766594648 Chr.5: 160569502-160569502 on Build GRCh38
rs906406797 Chr.5: 160569441-160569441 on Build GRCh38
rs780453264 Chr.5: 160569464-160569464 on Build GRCh38
rs758781976 Chr.5: 160569418-160569418 on Build GRCh38
rs771565170 Chr.5: 160569476-160569476 on Build GRCh38
rs888272576 Chr.5: 160569349-160569349 on Build GRCh38
rs771750402 Chr.5: 160569486-160569486 on Build GRCh38
rs749768097 Chr.5: 160569470-160569470 on Build GRCh38
rs760498668 Chr.5: 160569490-160569490 on Build GRCh38
rs773939732 Chr.5: 160569481-160569481 on Build GRCh38
rs932373590 Chr.5: 160569313-160569313 on Build GRCh38
rs752144547 Chr.5: 160569477-160569477 on Build GRCh38
rs755414630 Chr.5: 160569487-160569487 on Build GRCh38
rs1003558971 Chr.5: 160569459-160569459 on Build GRCh38
rs1039427229 Chr.5: 160569472-160569472 on Build GRCh38
rs1022451557 Chr.5: 160569491-160569491 on Build GRCh38
rs574290824 Chr.5: 160569330-160569330 on Build GRCh38
rs7709677 Chr.5: 160569383-160569383 on Build GRCh38
rs781329401 Chr.5: 160569447-160569447 on Build GRCh38
rs748544246 Chr.5: 160569449-160569449 on Build GRCh38
rs765554802 Chr.5: 160569377-160569377 on Build GRCh38

View Full Product Details