Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74573419-74573658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH3
Entrez Gene ID
127254
Gene Name
glutamate rich 3
Gene Aliases
C1orf173
UniGene
Hs.531182
Gene Chromosome Location
Chr.1: 74568111-74674343 on Build GRCh38
Transcripts
NM_001002912.4 XM_017000274.1 XM_017000275.1 XM_017000276.1 XM_017000277.1 XM_017000278.1 XM_017000279.1 XM_017000280.1 XM_017000281.1 XM_017000282.1 XM_017000283.1 XM_017000284.1 XM_017000285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344275 c.2231A>G p.N744S Chr.1: 74573479-74573479 on Build GRCh38
5677029 c.2277C>G p.S759S Chr.1: 74573433-74573433 on Build GRCh38
912095 c.2235C>A p.F745L Chr.1: 74573475-74573475 on Build GRCh38
1718218 c.2245G>A p.E749K Chr.1: 74573465-74573465 on Build GRCh38
5464110 c.2230A>C p.N744H Chr.1: 74573480-74573480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757092911 Chr.1: 74573516-74573516 on Build GRCh38
rs565631020 Chr.1: 74573489-74573489 on Build GRCh38
rs868480723 Chr.1: 74573604-74573604 on Build GRCh38
rs1005356911 Chr.1: 74573619-74573619 on Build GRCh38
rs992965028 Chr.1: 74573587-74573587 on Build GRCh38
rs549181694 Chr.1: 74573617-74573617 on Build GRCh38
rs77910454 Chr.1: 74573535-74573535 on Build GRCh38
rs779104109 Chr.1: 74573509-74573509 on Build GRCh38
rs924678680 Chr.1: 74573552-74573552 on Build GRCh38
rs762535200 Chr.1: 74573475-74573475 on Build GRCh38
rs888101750 Chr.1: 74573622-74573622 on Build GRCh38
rs750711491 Chr.1: 74573449-74573449 on Build GRCh38
rs916960196 Chr.1: 74573566-74573566 on Build GRCh38
rs371120806 Chr.1: 74573515-74573515 on Build GRCh38
rs1048128765 Chr.1: 74573607-74573607 on Build GRCh38
rs755890279 Chr.1: 74573543-74573543 on Build GRCh38
rs770302286 Chr.1: 74573566-74573567 on Build GRCh38
rs537266111 Chr.1: 74573591-74573591 on Build GRCh38
rs148986005 Chr.1: 74573441-74573441 on Build GRCh38
rs948931893 Chr.1: 74573598-74573598 on Build GRCh38
rs375493438 Chr.1: 74573491-74573491 on Build GRCh38
rs781196223 Chr.1: 74573534-74573534 on Build GRCh38
rs768094865 Chr.1: 74573448-74573448 on Build GRCh38
rs766647500 Chr.1: 74573474-74573474 on Build GRCh38
rs187348953 Chr.1: 74573582-74573582 on Build GRCh38
rs1057182089 Chr.1: 74573563-74573563 on Build GRCh38
rs372328125 Chr.1: 74573577-74573578 on Build GRCh38
rs762000632 Chr.1: 74573614-74573614 on Build GRCh38

View Full Product Details