Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186539166-186539439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYGS
Entrez Gene ID
1427
Gene Name
crystallin gamma S
Gene Aliases
CRYG8, CTRCT20
UniGene
Hs.376209
Gene Chromosome Location
Chr.3: 186538443-186544378 on Build GRCh38
Transcripts
NM_017541.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6407627 c.188C>T p.P63L Chr.3: 186539431-186539431 on Build GRCh38
6303710 c.254C>A p.A85D Chr.3: 186539365-186539365 on Build GRCh38
3590983 c.245C>T p.S82F Chr.3: 186539374-186539374 on Build GRCh38
243861 c.235C>T p.R79C Chr.3: 186539384-186539384 on Build GRCh38
7144033 c.245C>G p.S82C Chr.3: 186539374-186539374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568263645 Chr.3: 186539318-186539318 on Build GRCh38
rs140997568 Chr.3: 186539360-186539360 on Build GRCh38
rs768279275 Chr.3: 186539360-186539360 on Build GRCh38
rs965902080 Chr.3: 186539299-186539299 on Build GRCh38
rs1021944645 Chr.3: 186539244-186539244 on Build GRCh38
rs764969189 Chr.3: 186539383-186539383 on Build GRCh38
rs749524236 Chr.3: 186539354-186539354 on Build GRCh38
rs776271006 Chr.3: 186539394-186539394 on Build GRCh38
rs928671942 Chr.3: 186539406-186539406 on Build GRCh38
rs377244223 Chr.3: 186539306-186539306 on Build GRCh38
rs773875763 Chr.3: 186539286-186539286 on Build GRCh38
rs750236806 Chr.3: 186539405-186539405 on Build GRCh38
rs772501303 Chr.3: 186539384-186539384 on Build GRCh38
rs771128011 Chr.3: 186539359-186539359 on Build GRCh38
rs201962666 Chr.3: 186539314-186539314 on Build GRCh38
rs759306588 Chr.3: 186539309-186539309 on Build GRCh38
rs528601689 Chr.3: 186539259-186539259 on Build GRCh38
rs746873054 Chr.3: 186539319-186539319 on Build GRCh38
rs373678419 Chr.3: 186539387-186539387 on Build GRCh38
rs982838217 Chr.3: 186539381-186539381 on Build GRCh38
rs144812537 Chr.3: 186539404-186539404 on Build GRCh38
rs369814169 Chr.3: 186539334-186539334 on Build GRCh38
rs758310514 Chr.3: 186539317-186539317 on Build GRCh38
rs1030643258 Chr.3: 186539217-186539217 on Build GRCh38
rs928101709 Chr.3: 186539370-186539370 on Build GRCh38
rs750232128 Chr.3: 186539316-186539316 on Build GRCh38
rs950614681 Chr.3: 186539324-186539324 on Build GRCh38
rs747261060 Chr.3: 186539323-186539323 on Build GRCh38
rs890193865 Chr.3: 186539211-186539211 on Build GRCh38
rs761628147 Chr.3: 186539395-186539395 on Build GRCh38
rs892016868 Chr.3: 186539229-186539229 on Build GRCh38
rs930035003 Chr.3: 186539373-186539373 on Build GRCh38
rs766300240 Chr.3: 186539218-186539218 on Build GRCh38
rs1017888004 Chr.3: 186539281-186539281 on Build GRCh38
rs753571891 Chr.3: 186539307-186539307 on Build GRCh38
rs760206816 Chr.3: 186539304-186539304 on Build GRCh38
rs999228075 Chr.3: 186539215-186539215 on Build GRCh38
rs753814945 Chr.3: 186539237-186539237 on Build GRCh38
rs535693663 Chr.3: 186539388-186539388 on Build GRCh38
rs763714261 Chr.3: 186539389-186539389 on Build GRCh38
rs116212798 Chr.3: 186539251-186539251 on Build GRCh38
rs758237494 Chr.3: 186539416-186539416 on Build GRCh38
rs756217318 Chr.3: 186539327-186539327 on Build GRCh38

View Full Product Details