Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6017821-6018082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIMP2
Entrez Gene ID
7965
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases
JTV-1, JTV1, P38
UniGene
Hs.301613
Gene Chromosome Location
Chr.7: 6009241-6023834 on Build GRCh38
Transcripts
NM_001326606.1 NM_001326607.1 NM_001326609.1 NM_001326611.1 NM_006303.3 XM_005249847.3 XM_005249848.2 XM_005249850.3 XM_006715772.2 XM_011515519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1451703 c.383C>T p.P128L Chr.7: 6017854-6017854 on Build GRCh38
5979324 c.442G>A p.V148I Chr.7: 6017913-6017913 on Build GRCh38
3881950 c.376G>A p.A126T Chr.7: 6017847-6017847 on Build GRCh38
747342 c.467C>T p.S156F Chr.7: 6017938-6017938 on Build GRCh38
1623016 c.537A>C p.Q179H Chr.7: 6018008-6018008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528137967 Chr.7: 6018055-6018055 on Build GRCh38
rs61735883 Chr.7: 6017933-6017933 on Build GRCh38
rs768732685 Chr.7: 6017952-6017952 on Build GRCh38
rs534008494 Chr.7: 6017924-6017924 on Build GRCh38
rs747974072 Chr.7: 6017854-6017854 on Build GRCh38
rs757057598 Chr.7: 6018002-6018002 on Build GRCh38
rs747381930 Chr.7: 6018057-6018057 on Build GRCh38
rs773768066 Chr.7: 6017968-6017968 on Build GRCh38
rs374110509 Chr.7: 6017951-6017951 on Build GRCh38
rs761480147 Chr.7: 6017988-6017989 on Build GRCh38
rs765640941 Chr.7: 6017923-6017923 on Build GRCh38
rs750912720 Chr.7: 6017845-6017845 on Build GRCh38
rs373372974 Chr.7: 6017850-6017850 on Build GRCh38
rs779079471 Chr.7: 6018022-6018022 on Build GRCh38
rs767955463 Chr.7: 6017899-6017899 on Build GRCh38
rs755018110 Chr.7: 6018016-6018016 on Build GRCh38
rs145256692 Chr.7: 6017846-6017846 on Build GRCh38
rs762654416 Chr.7: 6017996-6017996 on Build GRCh38
rs373689127 Chr.7: 6017957-6017957 on Build GRCh38
rs931014900 Chr.7: 6017948-6017948 on Build GRCh38
rs549852720 Chr.7: 6017913-6017913 on Build GRCh38
rs17855441 Chr.7: 6017857-6017857 on Build GRCh38
rs754644786 Chr.7: 6017852-6017852 on Build GRCh38
rs746796665 Chr.7: 6017938-6017938 on Build GRCh38
rs774693908 Chr.7: 6017891-6017891 on Build GRCh38
rs141537839 Chr.7: 6017980-6017980 on Build GRCh38
rs929422782 Chr.7: 6018050-6018050 on Build GRCh38
rs771672642 Chr.7: 6017991-6017996 on Build GRCh38
rs774926931 Chr.7: 6017840-6017840 on Build GRCh38
rs751354172 Chr.7: 6017999-6017999 on Build GRCh38
rs750336358 Chr.7: 6018006-6018006 on Build GRCh38
rs761095840 Chr.7: 6017969-6017969 on Build GRCh38
rs34525431 Chr.7: 6017967-6017967 on Build GRCh38
rs758162658 Chr.7: 6017855-6017855 on Build GRCh38
rs774747119 Chr.7: 6017953-6017953 on Build GRCh38
rs1049065707 Chr.7: 6017893-6017893 on Build GRCh38
rs762290684 Chr.7: 6017894-6017894 on Build GRCh38
rs550952758 Chr.7: 6017847-6017847 on Build GRCh38
rs75895975 Chr.7: 6017932-6017932 on Build GRCh38
rs545201082 Chr.7: 6017939-6017939 on Build GRCh38
rs771380350 Chr.7: 6017971-6017971 on Build GRCh38
rs762237456 Chr.7: 6017843-6017843 on Build GRCh38
rs765096358 Chr.7: 6017884-6017884 on Build GRCh38
rs1013331223 Chr.7: 6017964-6017964 on Build GRCh38
rs554964581 Chr.7: 6017900-6017900 on Build GRCh38
rs778622819 Chr.7: 6017853-6017853 on Build GRCh38
rs777707483 Chr.7: 6017860-6017860 on Build GRCh38
rs370990323 Chr.7: 6017942-6017942 on Build GRCh38
rs777169814 Chr.7: 6017975-6017975 on Build GRCh38
rs748222802 Chr.7: 6018043-6018043 on Build GRCh38
rs773768396 Chr.7: 6017883-6017884 on Build GRCh38
rs377624555 Chr.7: 6018037-6018037 on Build GRCh38
rs542587504 Chr.7: 6017994-6017994 on Build GRCh38
rs946176280 Chr.7: 6017890-6017890 on Build GRCh38
rs761028688 Chr.7: 6017912-6017912 on Build GRCh38
rs764843355 Chr.7: 6017928-6017928 on Build GRCh38
rs746905456 Chr.7: 6017869-6017869 on Build GRCh38
rs778108674 Chr.7: 6018056-6018056 on Build GRCh38
rs374990034 Chr.7: 6017935-6017935 on Build GRCh38
rs768148274 Chr.7: 6017855-6017856 on Build GRCh38
rs957576738 Chr.7: 6018049-6018049 on Build GRCh38
rs766723249 Chr.7: 6018004-6018004 on Build GRCh38
rs200566166 Chr.7: 6018003-6018003 on Build GRCh38
rs137867185 Chr.7: 6017873-6017873 on Build GRCh38
rs769933568 Chr.7: 6017872-6017872 on Build GRCh38
rs560844337 Chr.7: 6017997-6017997 on Build GRCh38
rs777031773 Chr.7: 6018058-6018058 on Build GRCh38
rs981257367 Chr.7: 6018011-6018011 on Build GRCh38
rs923027283 Chr.7: 6017990-6017990 on Build GRCh38
rs536225300 Chr.7: 6017883-6017883 on Build GRCh38

View Full Product Details