Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157697193-157697456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
UniGene
Hs.292449
Gene Chromosome Location
Chr.1: 157676481-157700985 on Build GRCh38
Transcripts
NM_001320333.1 NM_052939.3 NR_135214.1 NR_135215.1 NR_135216.1 NR_135217.1 XM_006711145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3863077 c.667C>T p.Q223* Chr.1: 157697317-157697317 on Build GRCh38
897567 c.567T>G p.F189L Chr.1: 157697417-157697417 on Build GRCh38
4421601 c.695G>T p.S232I Chr.1: 157697289-157697289 on Build GRCh38
3475796 c.656G>T p.R219M Chr.1: 157697328-157697328 on Build GRCh38
7309495 c.749T>C p.M250T Chr.1: 157697235-157697235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746657472 Chr.1: 157697282-157697282 on Build GRCh38
rs200028494 Chr.1: 157697358-157697358 on Build GRCh38
rs768114034 Chr.1: 157697283-157697283 on Build GRCh38
rs777897301 Chr.1: 157697262-157697262 on Build GRCh38
rs757751434 Chr.1: 157697272-157697272 on Build GRCh38
rs368948019 Chr.1: 157697387-157697387 on Build GRCh38
rs754618197 Chr.1: 157697398-157697398 on Build GRCh38
rs202180859 Chr.1: 157697384-157697384 on Build GRCh38
rs747654707 Chr.1: 157697355-157697355 on Build GRCh38
rs750794333 Chr.1: 157697322-157697322 on Build GRCh38
rs751897935 Chr.1: 157697393-157697393 on Build GRCh38
rs771747301 Chr.1: 157697280-157697280 on Build GRCh38
rs138227125 Chr.1: 157697410-157697410 on Build GRCh38
rs756296436 Chr.1: 157697378-157697378 on Build GRCh38
rs757788314 Chr.1: 157697318-157697318 on Build GRCh38
rs201082888 Chr.1: 157697250-157697250 on Build GRCh38
rs762969025 Chr.1: 157697362-157697362 on Build GRCh38
rs375297389 Chr.1: 157697385-157697385 on Build GRCh38
rs750217294 Chr.1: 157697229-157697229 on Build GRCh38
rs764120395 Chr.1: 157697311-157697311 on Build GRCh38
rs187804313 Chr.1: 157697379-157697379 on Build GRCh38
rs753932075 Chr.1: 157697315-157697315 on Build GRCh38
rs745616221 Chr.1: 157697226-157697229 on Build GRCh38
rs758707560 Chr.1: 157697324-157697324 on Build GRCh38
rs755272086 Chr.1: 157697232-157697232 on Build GRCh38
rs75038928 Chr.1: 157697298-157697298 on Build GRCh38
rs761802836 Chr.1: 157697222-157697222 on Build GRCh38
rs760947770 Chr.1: 157697308-157697308 on Build GRCh38
rs773223997 Chr.1: 157697360-157697360 on Build GRCh38
rs752828046 Chr.1: 157697244-157697244 on Build GRCh38
rs767807516 Chr.1: 157697235-157697235 on Build GRCh38
rs77395825 Chr.1: 157697400-157697400 on Build GRCh38
rs769483978 Chr.1: 157697416-157697416 on Build GRCh38
rs760856382 Chr.1: 157697367-157697367 on Build GRCh38
rs776173025 Chr.1: 157697287-157697287 on Build GRCh38
rs768328117 Chr.1: 157697335-157697335 on Build GRCh38
rs775622973 Chr.1: 157697280-157697281 on Build GRCh38
rs149249986 Chr.1: 157697293-157697293 on Build GRCh38
rs987716008 Chr.1: 157697419-157697419 on Build GRCh38
rs774166384 Chr.1: 157697366-157697366 on Build GRCh38
rs770917354 Chr.1: 157697365-157697365 on Build GRCh38
rs78284410 Chr.1: 157697423-157697423 on Build GRCh38
rs752922608 Chr.1: 157697305-157697305 on Build GRCh38
rs772299241 Chr.1: 157697254-157697255 on Build GRCh38
rs746185833 Chr.1: 157697279-157697279 on Build GRCh38
rs575879706 Chr.1: 157697328-157697328 on Build GRCh38
rs779356254 Chr.1: 157697319-157697319 on Build GRCh38
rs760882805 Chr.1: 157697396-157697396 on Build GRCh38
rs776487151 Chr.1: 157697220-157697220 on Build GRCh38
rs775060165 Chr.1: 157697281-157697281 on Build GRCh38
rs559562792 Chr.1: 157697304-157697304 on Build GRCh38
rs761701291 Chr.1: 157697290-157697290 on Build GRCh38
rs761816945 Chr.1: 157697383-157697383 on Build GRCh38
rs577747260 Chr.1: 157697341-157697341 on Build GRCh38
rs746677031 Chr.1: 157697330-157697330 on Build GRCh38
rs765159800 Chr.1: 157697226-157697226 on Build GRCh38
rs764332398 Chr.1: 157697368-157697368 on Build GRCh38
rs749279862 Chr.1: 157697429-157697429 on Build GRCh38
rs768943016 Chr.1: 157697269-157697269 on Build GRCh38
rs373477619 Chr.1: 157697278-157697278 on Build GRCh38
rs769857512 Chr.1: 157697357-157697357 on Build GRCh38

View Full Product Details