Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8607134-8607363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPZ
Entrez Gene ID
8532
Gene Name
carboxypeptidase Z
Gene Aliases
-
UniGene
Hs.61995
Gene Chromosome Location
Chr.4: 8592660-8619761 on Build GRCh38
Transcripts
NM_001014447.2 NM_001014448.2 NM_003652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3726875 c.1138C>T p.H380Y Chr.4: 8607336-8607336 on Build GRCh38
3606734 c.1165C>T p.P389S Chr.4: 8607363-8607363 on Build GRCh38
74254 c.1069G>T p.V357L Chr.4: 8607267-8607267 on Build GRCh38
6347874 c.1072G>T p.A358S Chr.4: 8607270-8607270 on Build GRCh38
3606733 c.1147G>A p.D383N Chr.4: 8607345-8607345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753825431 Chr.4: 8607307-8607307 on Build GRCh38
rs933754191 Chr.4: 8607342-8607342 on Build GRCh38
rs749461918 Chr.4: 8607281-8607281 on Build GRCh38
rs878901264 Chr.4: 8607179-8607179 on Build GRCh38
rs991264856 Chr.4: 8607246-8607246 on Build GRCh38
rs745619644 Chr.4: 8607276-8607276 on Build GRCh38
rs748182777 Chr.4: 8607345-8607345 on Build GRCh38
rs774551989 Chr.4: 8607254-8607254 on Build GRCh38
rs745695442 Chr.4: 8607242-8607242 on Build GRCh38
rs890074453 Chr.4: 8607187-8607187 on Build GRCh38
rs765332088 Chr.4: 8607323-8607323 on Build GRCh38
rs374697973 Chr.4: 8607215-8607215 on Build GRCh38
rs372862610 Chr.4: 8607216-8607216 on Build GRCh38
rs775607598 Chr.4: 8607245-8607245 on Build GRCh38
rs945517763 Chr.4: 8607343-8607343 on Build GRCh38
rs569997837 Chr.4: 8607273-8607273 on Build GRCh38
rs768055195 Chr.4: 8607259-8607259 on Build GRCh38
rs370426454 Chr.4: 8607232-8607232 on Build GRCh38
rs1011096317 Chr.4: 8607188-8607188 on Build GRCh38
rs112822922 Chr.4: 8607314-8607314 on Build GRCh38
rs200800800 Chr.4: 8607263-8607263 on Build GRCh38
rs549814123 Chr.4: 8607212-8607212 on Build GRCh38
rs371681610 Chr.4: 8607312-8607312 on Build GRCh38
rs778477174 Chr.4: 8607224-8607224 on Build GRCh38
rs372440409 Chr.4: 8607266-8607266 on Build GRCh38
rs142705871 Chr.4: 8607344-8607344 on Build GRCh38
rs917007831 Chr.4: 8607163-8607163 on Build GRCh38
rs576914971 Chr.4: 8607182-8607182 on Build GRCh38
rs199933402 Chr.4: 8607328-8607328 on Build GRCh38
rs539972460 Chr.4: 8607243-8607243 on Build GRCh38
rs762529615 Chr.4: 8607158-8607158 on Build GRCh38
rs766662888 Chr.4: 8607301-8607301 on Build GRCh38
rs866318508 Chr.4: 8607302-8607302 on Build GRCh38
rs750399515 Chr.4: 8607336-8607336 on Build GRCh38
rs1054413383 Chr.4: 8607337-8607337 on Build GRCh38
rs761030283 Chr.4: 8607299-8607299 on Build GRCh38
rs1009745026 Chr.4: 8607222-8607222 on Build GRCh38
rs763023404 Chr.4: 8607251-8607251 on Build GRCh38
rs769481357 Chr.4: 8607279-8607279 on Build GRCh38
rs948364712 Chr.4: 8607176-8607176 on Build GRCh38
rs565373818 Chr.4: 8607217-8607217 on Build GRCh38
rs757394425 Chr.4: 8607331-8607332 on Build GRCh38
rs771049806 Chr.4: 8607240-8607240 on Build GRCh38
rs2302578 Chr.4: 8607252-8607252 on Build GRCh38
rs138876879 Chr.4: 8607320-8607320 on Build GRCh38
rs368809801 Chr.4: 8607260-8607260 on Build GRCh38
rs754597074 Chr.4: 8607220-8607220 on Build GRCh38
rs1044062780 Chr.4: 8607183-8607183 on Build GRCh38
rs757867469 Chr.4: 8607269-8607269 on Build GRCh38
rs560895990 Chr.4: 8607184-8607184 on Build GRCh38
rs2302579 Chr.4: 8607207-8607207 on Build GRCh38
rs768321224 Chr.4: 8607159-8607159 on Build GRCh38
rs773736737 Chr.4: 8607296-8607296 on Build GRCh38
rs936957370 Chr.4: 8607206-8607206 on Build GRCh38
rs141998267 Chr.4: 8607339-8607339 on Build GRCh38
rs371852436 Chr.4: 8607237-8607237 on Build GRCh38
rs748278819 Chr.4: 8607288-8607288 on Build GRCh38
rs772516941 Chr.4: 8607294-8607294 on Build GRCh38
rs762508406 Chr.4: 8607310-8607310 on Build GRCh38
rs768992436 Chr.4: 8607285-8607285 on Build GRCh38
rs146301697 Chr.4: 8607341-8607341 on Build GRCh38
rs746957189 Chr.4: 8607239-8607239 on Build GRCh38
rs780265772 Chr.4: 8607280-8607280 on Build GRCh38
rs548623673 Chr.4: 8607272-8607272 on Build GRCh38
rs774102876 Chr.4: 8607168-8607168 on Build GRCh38
rs866353388 Chr.4: 8607313-8607313 on Build GRCh38
rs367807288 Chr.4: 8607275-8607275 on Build GRCh38
rs752199686 Chr.4: 8607231-8607231 on Build GRCh38
rs756861411 Chr.4: 8607321-8607321 on Build GRCh38
rs761158013 Chr.4: 8607304-8607304 on Build GRCh38
rs376732104 Chr.4: 8607274-8607274 on Build GRCh38
rs939796365 Chr.4: 8607290-8607290 on Build GRCh38
rs537180657 Chr.4: 8607286-8607286 on Build GRCh38

View Full Product Details