Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 117819666-117819891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX18
Entrez Gene ID
8886
Gene Name
DEAD-box helicase 18
Gene Aliases
MrDb
UniGene
Hs.744922
Gene Chromosome Location
Chr.2: 117814679-117832377 on Build GRCh38
Transcripts
NM_006773.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5978440 c.396C>G p.N132K Chr.2: 117819674-117819674 on Build GRCh38
1305584 c.472G>A p.E158K Chr.2: 117819750-117819750 on Build GRCh38
6291409 c.480G>C p.E160D Chr.2: 117819758-117819758 on Build GRCh38
3836670 c.441A>G p.T147T Chr.2: 117819719-117819719 on Build GRCh38
6263412 c.403A>G p.K135E Chr.2: 117819681-117819681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373454164 Chr.2: 117819844-117819844 on Build GRCh38
rs776209040 Chr.2: 117819704-117819704 on Build GRCh38
rs372599758 Chr.2: 117819842-117819842 on Build GRCh38
rs763375289 Chr.2: 117819741-117819741 on Build GRCh38
rs145263993 Chr.2: 117819802-117819802 on Build GRCh38
rs1821332 Chr.2: 117819798-117819798 on Build GRCh38
rs747782932 Chr.2: 117819707-117819707 on Build GRCh38
rs752875674 Chr.2: 117819751-117819751 on Build GRCh38
rs773020420 Chr.2: 117819722-117819722 on Build GRCh38
rs897310314 Chr.2: 117819700-117819700 on Build GRCh38
rs749133358 Chr.2: 117819812-117819812 on Build GRCh38
rs912224564 Chr.2: 117819826-117819826 on Build GRCh38
rs985166420 Chr.2: 117819823-117819823 on Build GRCh38
rs368103045 Chr.2: 117819728-117819728 on Build GRCh38
rs200434046 Chr.2: 117819770-117819770 on Build GRCh38
rs867584752 Chr.2: 117819781-117819781 on Build GRCh38
rs772046319 Chr.2: 117819839-117819839 on Build GRCh38
rs897333970 Chr.2: 117819850-117819850 on Build GRCh38
rs572198569 Chr.2: 117819790-117819790 on Build GRCh38
rs369891514 Chr.2: 117819738-117819738 on Build GRCh38
rs143106122 Chr.2: 117819736-117819736 on Build GRCh38
rs754180245 Chr.2: 117819766-117819766 on Build GRCh38
rs1026291107 Chr.2: 117819764-117819764 on Build GRCh38
rs944319622 Chr.2: 117819835-117819835 on Build GRCh38
rs368855632 Chr.2: 117819825-117819825 on Build GRCh38
rs767299026 Chr.2: 117819742-117819744 on Build GRCh38
rs372103129 Chr.2: 117819797-117819797 on Build GRCh38
rs780687052 Chr.2: 117819796-117819796 on Build GRCh38
rs143461429 Chr.2: 117819714-117819714 on Build GRCh38
rs779466280 Chr.2: 117819775-117819775 on Build GRCh38
rs759487258 Chr.2: 117819821-117819821 on Build GRCh38
rs183530887 Chr.2: 117819862-117819862 on Build GRCh38
rs759755658 Chr.2: 117819742-117819742 on Build GRCh38
rs767691365 Chr.2: 117819744-117819744 on Build GRCh38
rs750919150 Chr.2: 117819776-117819776 on Build GRCh38
rs754502558 Chr.2: 117819779-117819779 on Build GRCh38
rs770843551 Chr.2: 117819814-117819814 on Build GRCh38
rs764394070 Chr.2: 117819765-117819765 on Build GRCh38
rs147371946 Chr.2: 117819702-117819702 on Build GRCh38
rs774487616 Chr.2: 117819820-117819820 on Build GRCh38
rs147612688 Chr.2: 117819864-117819864 on Build GRCh38
rs757592465 Chr.2: 117819771-117819771 on Build GRCh38
rs41279754 Chr.2: 117819701-117819701 on Build GRCh38
rs528106346 Chr.2: 117819756-117819756 on Build GRCh38

View Full Product Details