Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53459580-53459819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTB1
Entrez Gene ID
63948
Gene Name
DMRT like family B with proline rich C-terminal 1
Gene Aliases
-
UniGene
Hs.131654
Gene Chromosome Location
Chr.1: 53459399-53467488 on Build GRCh38
Transcripts
NM_033067.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748599 c.268G>A p.V90I Chr.1: 53459721-53459721 on Build GRCh38
4632663 c.249C>A p.A83A Chr.1: 53459702-53459702 on Build GRCh38
6203796 c.239C>T p.A80V Chr.1: 53459692-53459692 on Build GRCh38
5730875 c.273G>C p.P91P Chr.1: 53459726-53459726 on Build GRCh38
5352173 c.247_248insCCGCCC p.P87_V88insAP Chr.1: 53459700-53459701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148945511 Chr.1: 53459604-53459604 on Build GRCh38
rs769863672 Chr.1: 53459612-53459612 on Build GRCh38
rs949279620 Chr.1: 53459753-53459753 on Build GRCh38
rs780782973 Chr.1: 53459608-53459608 on Build GRCh38
rs772353431 Chr.1: 53459727-53459727 on Build GRCh38
rs1013817384 Chr.1: 53459651-53459651 on Build GRCh38
rs745730044 Chr.1: 53459610-53459610 on Build GRCh38
rs748254618 Chr.1: 53459717-53459717 on Build GRCh38
rs866444668 Chr.1: 53459632-53459632 on Build GRCh38
rs183181300 Chr.1: 53459683-53459683 on Build GRCh38
rs780194712 Chr.1: 53459615-53459615 on Build GRCh38
rs774418284 Chr.1: 53459622-53459622 on Build GRCh38
rs978070351 Chr.1: 53459711-53459716 on Build GRCh38
rs575081928 Chr.1: 53459700-53459701 on Build GRCh38
rs765656172 Chr.1: 53459654-53459654 on Build GRCh38
rs576813564 Chr.1: 53459739-53459739 on Build GRCh38
rs754214702 Chr.1: 53459597-53459597 on Build GRCh38
rs936609114 Chr.1: 53459730-53459730 on Build GRCh38
rs916468382 Chr.1: 53459752-53459752 on Build GRCh38
rs369311808 Chr.1: 53459603-53459603 on Build GRCh38
rs773481553 Chr.1: 53459729-53459729 on Build GRCh38
rs759796384 Chr.1: 53459653-53459653 on Build GRCh38
rs993839065 Chr.1: 53459670-53459670 on Build GRCh38
rs772007560 Chr.1: 53459625-53459625 on Build GRCh38
rs755756243 Chr.1: 53459674-53459674 on Build GRCh38
rs755437671 Chr.1: 53459598-53459598 on Build GRCh38
rs749298992 Chr.1: 53459688-53459688 on Build GRCh38
rs1026618415 Chr.1: 53459672-53459672 on Build GRCh38
rs754382337 Chr.1: 53459637-53459637 on Build GRCh38
rs749094517 Chr.1: 53459619-53459619 on Build GRCh38
rs768540318 Chr.1: 53459620-53459620 on Build GRCh38
rs1018256590 Chr.1: 53459697-53459699 on Build GRCh38
rs773286484 Chr.1: 53459633-53459633 on Build GRCh38
rs558152882 Chr.1: 53459700-53459700 on Build GRCh38
rs1046618786 Chr.1: 53459790-53459790 on Build GRCh38
rs923418672 Chr.1: 53459705-53459706 on Build GRCh38
rs976922588 Chr.1: 53459601-53459601 on Build GRCh38
rs372787995 Chr.1: 53459634-53459634 on Build GRCh38
rs376953028 Chr.1: 53459659-53459659 on Build GRCh38
rs771048281 Chr.1: 53459746-53459746 on Build GRCh38
rs201518564 Chr.1: 53459662-53459662 on Build GRCh38
rs757031376 Chr.1: 53459637-53459639 on Build GRCh38
rs746877148 Chr.1: 53459710-53459711 on Build GRCh38
rs766482978 Chr.1: 53459706-53459707 on Build GRCh38
rs777072132 Chr.1: 53459766-53459766 on Build GRCh38
rs768289459 Chr.1: 53459710-53459710 on Build GRCh38
rs186491868 Chr.1: 53459716-53459716 on Build GRCh38
rs201603810 Chr.1: 53459636-53459636 on Build GRCh38
rs778707127 Chr.1: 53459644-53459646 on Build GRCh38
rs771736545 Chr.1: 53459701-53459706 on Build GRCh38
rs749935479 Chr.1: 53459671-53459671 on Build GRCh38
rs894953749 Chr.1: 53459643-53459643 on Build GRCh38
rs768785806 Chr.1: 53459722-53459723 on Build GRCh38
rs756670189 Chr.1: 53459607-53459607 on Build GRCh38
rs780788164 Chr.1: 53459664-53459664 on Build GRCh38

View Full Product Details