Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76351635-76351894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC158
Entrez Gene ID
339965
Gene Name
coiled-coil domain containing 158
Gene Aliases
-
UniGene
Hs.529680
Gene Chromosome Location
Chr.4: 76313039-76421920 on Build GRCh38
Transcripts
NM_001042784.1 XM_005262974.4 XM_006714204.3 XM_011531908.2 XM_011531909.2 XM_011531910.2 XM_011531911.1 XM_011531912.2 XM_011531913.1 XM_011531914.2 XM_011531915.2 XM_011531916.2 XM_011531917.1 XM_017008160.1 XM_017008161.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2832210 c.2525C>A p.T842N Chr.4: 76351733-76351733 on Build GRCh38
1328398 c.2509T>A p.L837I Chr.4: 76351749-76351749 on Build GRCh38
149699 c.2486G>A p.R829H Chr.4: 76351772-76351772 on Build GRCh38
5644337 c.2487T>C p.R829R Chr.4: 76351771-76351771 on Build GRCh38
319273 c.2523C>G p.H841Q Chr.4: 76351735-76351735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751983822 Chr.4: 76351707-76351707 on Build GRCh38
rs566541281 Chr.4: 76351723-76351723 on Build GRCh38
rs771984459 Chr.4: 76351847-76351847 on Build GRCh38
rs765281468 Chr.4: 76351700-76351700 on Build GRCh38
rs768108011 Chr.4: 76351835-76351835 on Build GRCh38
rs769141931 Chr.4: 76351675-76351675 on Build GRCh38
rs781018971 Chr.4: 76351724-76351724 on Build GRCh38
rs759733116 Chr.4: 76351770-76351770 on Build GRCh38
rs112905571 Chr.4: 76351678-76351678 on Build GRCh38
rs772946405 Chr.4: 76351849-76351849 on Build GRCh38
rs553105680 Chr.4: 76351768-76351768 on Build GRCh38
rs771814803 Chr.4: 76351754-76351754 on Build GRCh38
rs752069785 Chr.4: 76351782-76351782 on Build GRCh38
rs377204647 Chr.4: 76351684-76351684 on Build GRCh38
rs149122809 Chr.4: 76351772-76351772 on Build GRCh38
rs745551156 Chr.4: 76351730-76351730 on Build GRCh38
rs371270141 Chr.4: 76351856-76351856 on Build GRCh38
rs757803416 Chr.4: 76351788-76351788 on Build GRCh38
rs375808480 Chr.4: 76351751-76351751 on Build GRCh38
rs752806941 Chr.4: 76351702-76351702 on Build GRCh38
rs781545404 Chr.4: 76351715-76351718 on Build GRCh38
rs774200495 Chr.4: 76351683-76351683 on Build GRCh38
rs374410757 Chr.4: 76351752-76351752 on Build GRCh38
rs375618007 Chr.4: 76351758-76351758 on Build GRCh38
rs781726058 Chr.4: 76351792-76351792 on Build GRCh38
rs372527938 Chr.4: 76351691-76351691 on Build GRCh38
rs764717970 Chr.4: 76351778-76351778 on Build GRCh38
rs890931536 Chr.4: 76351714-76351714 on Build GRCh38
rs778854824 Chr.4: 76351842-76351842 on Build GRCh38
rs750790737 Chr.4: 76351793-76351793 on Build GRCh38
rs758892404 Chr.4: 76351704-76351704 on Build GRCh38
rs779692728 Chr.4: 76351813-76351813 on Build GRCh38
rs779490412 Chr.4: 76351737-76351737 on Build GRCh38
rs201357214 Chr.4: 76351733-76351733 on Build GRCh38
rs372793788 Chr.4: 76351680-76351680 on Build GRCh38
rs770347124 Chr.4: 76351791-76351791 on Build GRCh38
rs759940666 Chr.4: 76351866-76351866 on Build GRCh38
rs755117006 Chr.4: 76351695-76351695 on Build GRCh38
rs756959462 Chr.4: 76351756-76351756 on Build GRCh38
rs909965143 Chr.4: 76351790-76351790 on Build GRCh38
rs368831343 Chr.4: 76351690-76351690 on Build GRCh38
rs183278785 Chr.4: 76351773-76351773 on Build GRCh38
rs201154183 Chr.4: 76351831-76351831 on Build GRCh38
rs760449795 Chr.4: 76351851-76351851 on Build GRCh38
rs755792610 Chr.4: 76351801-76351801 on Build GRCh38
rs538470259 Chr.4: 76351826-76351826 on Build GRCh38
rs778355673 Chr.4: 76351705-76351705 on Build GRCh38
rs748511909 Chr.4: 76351779-76351780 on Build GRCh38

View Full Product Details