Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105531935-105532160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001627.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3584843 c.438C>T p.L146L Chr.3: 105532045-105532045 on Build GRCh38
2916407 c.453delA p.K153fs*34 Chr.3: 105532060-105532060 on Build GRCh38
7259266 c.452_453insA p.L154fs*3 Chr.3: 105532059-105532060 on Build GRCh38
1742743 c.458A>T p.K153M Chr.3: 105532065-105532065 on Build GRCh38
5397712 c.398A>G p.Q133R Chr.3: 105532005-105532005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530196302 Chr.3: 105532040-105532040 on Build GRCh38
rs199708955 Chr.3: 105532070-105532070 on Build GRCh38
rs747658659 Chr.3: 105531990-105531990 on Build GRCh38
rs990855414 Chr.3: 105532029-105532029 on Build GRCh38
rs746683074 Chr.3: 105532036-105532036 on Build GRCh38
rs371767685 Chr.3: 105532060-105532060 on Build GRCh38
rs961248832 Chr.3: 105532016-105532016 on Build GRCh38
rs745734770 Chr.3: 105532045-105532045 on Build GRCh38
rs566395475 Chr.3: 105532033-105532033 on Build GRCh38
rs773441884 Chr.3: 105532081-105532081 on Build GRCh38
rs754321301 Chr.3: 105531973-105531973 on Build GRCh38
rs373229018 Chr.3: 105532025-105532025 on Build GRCh38
rs766708280 Chr.3: 105532087-105532087 on Build GRCh38
rs756467823 Chr.3: 105531967-105531967 on Build GRCh38
rs769710275 Chr.3: 105532055-105532055 on Build GRCh38
rs767532712 Chr.3: 105532076-105532076 on Build GRCh38
rs77939390 Chr.3: 105532116-105532116 on Build GRCh38
rs755556864 Chr.3: 105531978-105531978 on Build GRCh38
rs548773073 Chr.3: 105532084-105532084 on Build GRCh38
rs370436000 Chr.3: 105531988-105531988 on Build GRCh38
rs928242011 Chr.3: 105532012-105532012 on Build GRCh38
rs766685490 Chr.3: 105531969-105531969 on Build GRCh38
rs750658744 Chr.3: 105531965-105531965 on Build GRCh38
rs201545016 Chr.3: 105532046-105532046 on Build GRCh38
rs144776789 Chr.3: 105532093-105532094 on Build GRCh38
rs754158436 Chr.3: 105532091-105532091 on Build GRCh38

View Full Product Details