Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28882285-28882554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT2
Entrez Gene ID
2319
Gene Name
flotillin 2
Gene Aliases
ECS-1, ECS1, ESA, ESA1, M17S1
UniGene
Hs.514038
Gene Chromosome Location
Chr.17: 28879339-28897728 on Build GRCh38
Transcripts
NM_004475.2 XM_005257950.3 XM_005257953.2 XM_017024394.1 XM_017024395.1 XM_017024396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5429432 c.506A>C p.K169T Chr.17: 28882410-28882410 on Build GRCh38
1610011 c.547G>A p.V183M Chr.17: 28882369-28882369 on Build GRCh38
2801105 c.577C>T p.R193W Chr.17: 28882339-28882339 on Build GRCh38
977040 c.519C>T p.A173A Chr.17: 28882397-28882397 on Build GRCh38
4065082 c.567C>T p.D189D Chr.17: 28882349-28882349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758713849 Chr.17: 28882438-28882438 on Build GRCh38
rs138029483 Chr.17: 28882407-28882407 on Build GRCh38
rs760793127 Chr.17: 28882493-28882493 on Build GRCh38
rs368057780 Chr.17: 28882456-28882456 on Build GRCh38
rs776108358 Chr.17: 28882354-28882354 on Build GRCh38
rs774268977 Chr.17: 28882394-28882394 on Build GRCh38
rs766671749 Chr.17: 28882496-28882496 on Build GRCh38
rs759256521 Chr.17: 28882364-28882364 on Build GRCh38
rs755011074 Chr.17: 28882526-28882526 on Build GRCh38
rs376860005 Chr.17: 28882396-28882396 on Build GRCh38
rs762982686 Chr.17: 28882374-28882374 on Build GRCh38
rs765323705 Chr.17: 28882534-28882534 on Build GRCh38
rs879730539 Chr.17: 28882338-28882338 on Build GRCh38
rs1034169326 Chr.17: 28882515-28882515 on Build GRCh38
rs200329095 Chr.17: 28882397-28882397 on Build GRCh38
rs747308471 Chr.17: 28882472-28882472 on Build GRCh38
rs748685439 Chr.17: 28882437-28882437 on Build GRCh38
rs775072847 Chr.17: 28882370-28882370 on Build GRCh38
rs373662636 Chr.17: 28882348-28882348 on Build GRCh38
rs778214135 Chr.17: 28882447-28882447 on Build GRCh38
rs769216547 Chr.17: 28882339-28882339 on Build GRCh38
rs774395363 Chr.17: 28882478-28882478 on Build GRCh38
rs770539405 Chr.17: 28882352-28882352 on Build GRCh38
rs769558391 Chr.17: 28882459-28882459 on Build GRCh38
rs750488943 Chr.17: 28882307-28882307 on Build GRCh38
rs753882638 Chr.17: 28882499-28882499 on Build GRCh38
rs572308228 Chr.17: 28882473-28882473 on Build GRCh38
rs779435564 Chr.17: 28882430-28882430 on Build GRCh38
rs1013505719 Chr.17: 28882433-28882433 on Build GRCh38
rs188812089 Chr.17: 28882492-28882492 on Build GRCh38
rs200313210 Chr.17: 28882454-28882454 on Build GRCh38
rs184928815 Chr.17: 28882426-28882426 on Build GRCh38
rs754006379 Chr.17: 28882415-28882415 on Build GRCh38
rs769129691 Chr.17: 28882369-28882369 on Build GRCh38
rs756094041 Chr.17: 28882317-28882317 on Build GRCh38
rs780192508 Chr.17: 28882333-28882333 on Build GRCh38
rs112013773 Chr.17: 28882453-28882453 on Build GRCh38
rs749688498 Chr.17: 28882336-28882336 on Build GRCh38
rs780181585 Chr.17: 28882471-28882490 on Build GRCh38
rs59926480 Chr.17: 28882406-28882406 on Build GRCh38
rs537594430 Chr.17: 28882349-28882349 on Build GRCh38
rs750874148 Chr.17: 28882400-28882400 on Build GRCh38
rs371970127 Chr.17: 28882464-28882464 on Build GRCh38
rs1046289766 Chr.17: 28882360-28882360 on Build GRCh38
rs200865799 Chr.17: 28882378-28882378 on Build GRCh38

View Full Product Details