Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98089686-98089939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH5
Entrez Gene ID
203111
Gene Name
glutamate rich 5
Gene Aliases
C8orf47
UniGene
Hs.171455
Gene Chromosome Location
Chr.8: 98064522-98093610 on Build GRCh38
Transcripts
NM_001170806.1 NM_173549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527828 c.894G>A p.Q298Q Chr.8: 98089911-98089911 on Build GRCh38
2792506 c.814G>C p.D272H Chr.8: 98089831-98089831 on Build GRCh38
1102871 c.846G>T p.M282I Chr.8: 98089863-98089863 on Build GRCh38
3902332 c.698G>C p.G233A Chr.8: 98089715-98089715 on Build GRCh38
1102870 c.779G>T p.R260I Chr.8: 98089796-98089796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190321700 Chr.8: 98089800-98089800 on Build GRCh38
rs561949961 Chr.8: 98089805-98089805 on Build GRCh38
rs999891963 Chr.8: 98089908-98089908 on Build GRCh38
rs754714227 Chr.8: 98089850-98089850 on Build GRCh38
rs144040008 Chr.8: 98089745-98089745 on Build GRCh38
rs201678120 Chr.8: 98089753-98089753 on Build GRCh38
rs772405768 Chr.8: 98089803-98089803 on Build GRCh38
rs772888103 Chr.8: 98089787-98089787 on Build GRCh38
rs550422485 Chr.8: 98089818-98089818 on Build GRCh38
rs765475482 Chr.8: 98089822-98089822 on Build GRCh38
rs775854894 Chr.8: 98089902-98089902 on Build GRCh38
rs770478991 Chr.8: 98089901-98089901 on Build GRCh38
rs764583969 Chr.8: 98089813-98089813 on Build GRCh38
rs938077711 Chr.8: 98089817-98089817 on Build GRCh38
rs747866229 Chr.8: 98089761-98089761 on Build GRCh38
rs747768336 Chr.8: 98089871-98089871 on Build GRCh38
rs777590424 Chr.8: 98089885-98089885 on Build GRCh38
rs570401772 Chr.8: 98089843-98089843 on Build GRCh38
rs780062697 Chr.8: 98089746-98089746 on Build GRCh38
rs754807565 Chr.8: 98089751-98089751 on Build GRCh38
rs373715169 Chr.8: 98089812-98089812 on Build GRCh38
rs765352544 Chr.8: 98089718-98089718 on Build GRCh38
rs372813375 Chr.8: 98089915-98089915 on Build GRCh38
rs1032660457 Chr.8: 98089909-98089909 on Build GRCh38
rs1056654614 Chr.8: 98089821-98089821 on Build GRCh38
rs904103433 Chr.8: 98089898-98089898 on Build GRCh38
rs149657750 Chr.8: 98089849-98089849 on Build GRCh38
rs750333521 Chr.8: 98089727-98089727 on Build GRCh38
rs546455247 Chr.8: 98089721-98089721 on Build GRCh38
rs11994440 Chr.8: 98089877-98089877 on Build GRCh38
rs35368005 Chr.8: 98089748-98089748 on Build GRCh38
rs769404309 Chr.8: 98089762-98089762 on Build GRCh38
rs140159231 Chr.8: 98089893-98089893 on Build GRCh38
rs145439382 Chr.8: 98089814-98089814 on Build GRCh38
rs781179086 Chr.8: 98089867-98089867 on Build GRCh38
rs1052500998 Chr.8: 98089856-98089856 on Build GRCh38
rs758522369 Chr.8: 98089846-98089846 on Build GRCh38

View Full Product Details