Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34996229-34996462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB5
Entrez Gene ID
25822
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
Hsc40
UniGene
Hs.237506
Gene Chromosome Location
Chr.9: 34989369-34998433 on Build GRCh38
Transcripts
NM_001135004.2 NM_001135005.2 NM_012266.5 XM_005251428.3 XM_006716751.2 XM_011517842.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5998543 c.706A>G p.S236G Chr.9: 34996417-34996417 on Build GRCh38
1569281 c.565G>A p.G189S Chr.9: 34996276-34996276 on Build GRCh38
1569282 c.223G>A p.G75S Chr.9: 34996276-34996276 on Build GRCh38
3907060 c.688C>T p.R230C Chr.9: 34996399-34996399 on Build GRCh38
6855436 c.623A>G p.D208G Chr.9: 34996460-34996460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770403825 Chr.9: 34996340-34996340 on Build GRCh38
rs376527177 Chr.9: 34996437-34996437 on Build GRCh38
rs777072091 Chr.9: 34996324-34996324 on Build GRCh38
rs752945019 Chr.9: 34996378-34996378 on Build GRCh38
rs748881964 Chr.9: 34996279-34996280 on Build GRCh38
rs536002468 Chr.9: 34996347-34996347 on Build GRCh38
rs1055753965 Chr.9: 34996355-34996355 on Build GRCh38
rs887163513 Chr.9: 34996438-34996438 on Build GRCh38
rs768445172 Chr.9: 34996282-34996285 on Build GRCh38
rs937578940 Chr.9: 34996320-34996320 on Build GRCh38
rs772171150 Chr.9: 34996276-34996276 on Build GRCh38
rs755790761 Chr.9: 34996251-34996253 on Build GRCh38
rs759139617 Chr.9: 34996390-34996390 on Build GRCh38
rs763996789 Chr.9: 34996406-34996406 on Build GRCh38
rs774096148 Chr.9: 34996292-34996300 on Build GRCh38
rs200305109 Chr.9: 34996398-34996398 on Build GRCh38
rs769040530 Chr.9: 34996319-34996319 on Build GRCh38
rs375297341 Chr.9: 34996291-34996291 on Build GRCh38
rs762320036 Chr.9: 34996329-34996329 on Build GRCh38
rs199764155 Chr.9: 34996409-34996409 on Build GRCh38
rs201082390 Chr.9: 34996422-34996422 on Build GRCh38
rs897796477 Chr.9: 34996431-34996431 on Build GRCh38
rs760373162 Chr.9: 34996279-34996279 on Build GRCh38
rs140560026 Chr.9: 34996429-34996429 on Build GRCh38
rs767050639 Chr.9: 34996399-34996399 on Build GRCh38
rs371839967 Chr.9: 34996417-34996417 on Build GRCh38
rs765696926 Chr.9: 34996275-34996275 on Build GRCh38
rs78312215 Chr.9: 34996433-34996433 on Build GRCh38
rs747342206 Chr.9: 34996286-34996286 on Build GRCh38
rs779745578 Chr.9: 34996260-34996260 on Build GRCh38
rs200844864 Chr.9: 34996400-34996400 on Build GRCh38

View Full Product Details