Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82583848-82584097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK2
Entrez Gene ID
3607
Gene Name
forkhead box K2
Gene Aliases
ILF, ILF-1, ILF1
UniGene
Hs.591140
Gene Chromosome Location
Chr.17: 82519718-82604607 on Build GRCh38
Transcripts
NM_004514.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882963 c.1104-1G>A p.? Chr.17: 82584012-82584012 on Build GRCh38
3523890 c.1167C>T p.T389T Chr.17: 82584076-82584076 on Build GRCh38
6228237 c.1128C>T p.H376H Chr.17: 82584037-82584037 on Build GRCh38
7146809 c.1176C>T p.S392S Chr.17: 82584085-82584085 on Build GRCh38
2806989 c.1129G>A p.A377T Chr.17: 82584038-82584038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771626203 Chr.17: 82583987-82583987 on Build GRCh38
rs760447310 Chr.17: 82584019-82584019 on Build GRCh38
rs367589541 Chr.17: 82583995-82583995 on Build GRCh38
rs759226071 Chr.17: 82583872-82583872 on Build GRCh38
rs1026543818 Chr.17: 82583942-82583942 on Build GRCh38
rs201121040 Chr.17: 82583984-82583984 on Build GRCh38
rs758615505 Chr.17: 82584042-82584042 on Build GRCh38
rs370530166 Chr.17: 82584003-82584003 on Build GRCh38
rs758596480 Chr.17: 82583971-82583971 on Build GRCh38
rs371456085 Chr.17: 82583973-82583973 on Build GRCh38
rs757703270 Chr.17: 82584067-82584067 on Build GRCh38
rs746403456 Chr.17: 82584076-82584076 on Build GRCh38
rs189873421 Chr.17: 82583877-82583877 on Build GRCh38
rs532399289 Chr.17: 82584000-82584000 on Build GRCh38
rs899295745 Chr.17: 82583898-82583898 on Build GRCh38
rs751776107 Chr.17: 82584061-82584061 on Build GRCh38
rs146692675 Chr.17: 82584037-82584037 on Build GRCh38
rs772942419 Chr.17: 82584017-82584017 on Build GRCh38
rs371667664 Chr.17: 82584039-82584039 on Build GRCh38
rs777292072 Chr.17: 82583990-82583990 on Build GRCh38
rs778044306 Chr.17: 82584047-82584047 on Build GRCh38
rs766101638 Chr.17: 82584033-82584033 on Build GRCh38
rs138125214 Chr.17: 82584068-82584068 on Build GRCh38
rs766997277 Chr.17: 82584038-82584038 on Build GRCh38

View Full Product Details