Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16391262-16391531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SZRD1
Entrez Gene ID
26099
Gene Name
SUZ RNA binding domain containing 1
Gene Aliases
C1orf144
UniGene
Hs.252967
Gene Chromosome Location
Chr.1: 16367030-16398148 on Build GRCh38
Transcripts
NM_001114600.2 NM_001271869.1 NR_073500.1 NR_073501.1 NR_073502.1 NR_073503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2185560 c.70_71insA p.L27fs*25 Chr.1: 16391393-16391394 on Build GRCh38
4185633 c.57_60delAGAC p.R21fs*6 Chr.1: 16391380-16391383 on Build GRCh38
181849 c.71delA p.K26fs*2 Chr.1: 16391394-16391394 on Build GRCh38
899058 c.65G>A p.R22Q Chr.1: 16391388-16391388 on Build GRCh38
6459572 c.54A>G p.E18E Chr.1: 16391377-16391377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941163072 Chr.1: 16391473-16391473 on Build GRCh38
rs998578450 Chr.1: 16391494-16391494 on Build GRCh38
rs902921215 Chr.1: 16391490-16391490 on Build GRCh38
rs972202992 Chr.1: 16391338-16391339 on Build GRCh38
rs186642606 Chr.1: 16391504-16391504 on Build GRCh38
rs1056690459 Chr.1: 16391348-16391348 on Build GRCh38
rs367782069 Chr.1: 16391318-16391319 on Build GRCh38
rs923447591 Chr.1: 16391339-16391339 on Build GRCh38
rs771805412 Chr.1: 16391440-16391440 on Build GRCh38
rs41302001 Chr.1: 16391476-16391476 on Build GRCh38
rs947045026 Chr.1: 16391474-16391474 on Build GRCh38
rs948429194 Chr.1: 16391377-16391377 on Build GRCh38
rs553807004 Chr.1: 16391317-16391318 on Build GRCh38
rs939607182 Chr.1: 16391310-16391310 on Build GRCh38
rs1056862239 Chr.1: 16391500-16391500 on Build GRCh38
rs889771189 Chr.1: 16391508-16391508 on Build GRCh38
rs933554701 Chr.1: 16391371-16391371 on Build GRCh38
rs538767701 Chr.1: 16391498-16391498 on Build GRCh38
rs1051121883 Chr.1: 16391380-16391383 on Build GRCh38
rs535720227 Chr.1: 16391485-16391485 on Build GRCh38
rs550436406 Chr.1: 16391383-16391383 on Build GRCh38
rs569752037 Chr.1: 16391406-16391406 on Build GRCh38
rs924162724 Chr.1: 16391286-16391286 on Build GRCh38
rs916905348 Chr.1: 16391359-16391359 on Build GRCh38
rs778883651 Chr.1: 16391400-16391400 on Build GRCh38
rs962145793 Chr.1: 16391319-16391319 on Build GRCh38
rs1029612108 Chr.1: 16391507-16391507 on Build GRCh38

View Full Product Details