Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73236777-73237030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf80
Entrez Gene ID
55028
Gene Name
chromosome 17 open reading frame 80
Gene Aliases
HLC-8, MIG3, SPEP1
UniGene
Hs.12929
Gene Chromosome Location
Chr.17: 73232233-73248959 on Build GRCh38
Transcripts
NM_001100621.2 NM_001100622.2 NM_001288770.1 NM_001288771.1 NM_017941.5 NR_110105.1 XM_005257487.3 XM_006721966.3 XM_011524961.1 XM_011524962.2 XM_017024806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5464187 c.1387G>T p.E463* Chr.17: 73236869-73236869 on Build GRCh38
6292384 c.1355G>T p.G452V Chr.17: 73236837-73236837 on Build GRCh38
5571540 c.1513G>A p.V505I Chr.17: 73236995-73236995 on Build GRCh38
3890306 c.1321C>T p.Q441* Chr.17: 73236803-73236803 on Build GRCh38
5658270 c.1479A>G p.Q493Q Chr.17: 73236961-73236961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964050565 Chr.17: 73236840-73236840 on Build GRCh38
rs138455388 Chr.17: 73236814-73236814 on Build GRCh38
rs112384237 Chr.17: 73236973-73236973 on Build GRCh38
rs768674849 Chr.17: 73236886-73236886 on Build GRCh38
rs755965159 Chr.17: 73236809-73236809 on Build GRCh38
rs776803820 Chr.17: 73236829-73236829 on Build GRCh38
rs776233311 Chr.17: 73236998-73236998 on Build GRCh38
rs762089285 Chr.17: 73236830-73236830 on Build GRCh38
rs770043007 Chr.17: 73236832-73236832 on Build GRCh38
rs781182841 Chr.17: 73236983-73236983 on Build GRCh38
rs147066698 Chr.17: 73236800-73236800 on Build GRCh38
rs372605437 Chr.17: 73236966-73236966 on Build GRCh38
rs750347726 Chr.17: 73236977-73236977 on Build GRCh38
rs781017192 Chr.17: 73236798-73236798 on Build GRCh38
rs748294541 Chr.17: 73236984-73236984 on Build GRCh38
rs76323272 Chr.17: 73236950-73236950 on Build GRCh38
rs568714232 Chr.17: 73236915-73236915 on Build GRCh38
rs753696035 Chr.17: 73236904-73236904 on Build GRCh38
rs202077581 Chr.17: 73236868-73236868 on Build GRCh38
rs61729639 Chr.17: 73236851-73236851 on Build GRCh38
rs368978948 Chr.17: 73236815-73236815 on Build GRCh38
rs752382573 Chr.17: 73236880-73236880 on Build GRCh38
rs753482726 Chr.17: 73236969-73236969 on Build GRCh38
rs771253294 Chr.17: 73236945-73236945 on Build GRCh38
rs117209041 Chr.17: 73236979-73236979 on Build GRCh38
rs766943500 Chr.17: 73236860-73236860 on Build GRCh38
rs76050886 Chr.17: 73236816-73236816 on Build GRCh38
rs201365165 Chr.17: 73236995-73236995 on Build GRCh38
rs755046193 Chr.17: 73236910-73236910 on Build GRCh38
rs760174353 Chr.17: 73236952-73236952 on Build GRCh38
rs777679556 Chr.17: 73236810-73236810 on Build GRCh38
rs868220287 Chr.17: 73236837-73236837 on Build GRCh38
rs377088562 Chr.17: 73236986-73236986 on Build GRCh38
rs779954924 Chr.17: 73236819-73236819 on Build GRCh38
rs192451785 Chr.17: 73236989-73236989 on Build GRCh38
rs759083212 Chr.17: 73236873-73236873 on Build GRCh38
rs556938635 Chr.17: 73236992-73236993 on Build GRCh38
rs767075816 Chr.17: 73236955-73236955 on Build GRCh38
rs780801472 Chr.17: 73236971-73236971 on Build GRCh38
rs11869253 Chr.17: 73236991-73236991 on Build GRCh38
rs763828300 Chr.17: 73236967-73236967 on Build GRCh38
rs773411008 Chr.17: 73236919-73236919 on Build GRCh38
rs964820192 Chr.17: 73236992-73236992 on Build GRCh38
rs747553563 Chr.17: 73236823-73236825 on Build GRCh38
rs763304786 Chr.17: 73236852-73236852 on Build GRCh38
rs770062408 Chr.17: 73236917-73236917 on Build GRCh38
rs745846965 Chr.17: 73236908-73236908 on Build GRCh38
rs775141494 Chr.17: 73236956-73236956 on Build GRCh38
rs373585783 Chr.17: 73236825-73236825 on Build GRCh38
rs778038391 Chr.17: 73236987-73236987 on Build GRCh38
rs376693946 Chr.17: 73236944-73236944 on Build GRCh38
rs747134947 Chr.17: 73236975-73236976 on Build GRCh38
rs757093143 Chr.17: 73236905-73236905 on Build GRCh38
rs867399298 Chr.17: 73236874-73236874 on Build GRCh38
rs754641328 Chr.17: 73236797-73236797 on Build GRCh38
rs894221671 Chr.17: 73236975-73236975 on Build GRCh38
rs755802112 Chr.17: 73236884-73236884 on Build GRCh38
rs747113349 Chr.17: 73236823-73236823 on Build GRCh38
rs778886756 Chr.17: 73236906-73236906 on Build GRCh38
rs771309156 Chr.17: 73236990-73236990 on Build GRCh38
rs765084585 Chr.17: 73236976-73236976 on Build GRCh38
rs763728803 Chr.17: 73236888-73236888 on Build GRCh38
rs977142143 Chr.17: 73236847-73236847 on Build GRCh38

View Full Product Details