Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99004819-99005092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL15A1
Entrez Gene ID
1306
Gene Name
collagen type XV alpha 1 chain
Gene Aliases
-
UniGene
Hs.409034
Gene Chromosome Location
Chr.9: 98943392-99070792 on Build GRCh38
Transcripts
NM_001855.4 XM_011518214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1458881 c.1315T>A p.S439T Chr.9: 99005012-99005012 on Build GRCh38
5721364 c.1322C>T p.S441F Chr.9: 99005019-99005019 on Build GRCh38
3763484 c.1336G>A p.G446R Chr.9: 99005033-99005033 on Build GRCh38
5459712 c.1324G>A p.A442T Chr.9: 99005021-99005021 on Build GRCh38
81034 c.1302G>A p.G434G Chr.9: 99004999-99004999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751001360 Chr.9: 99005041-99005041 on Build GRCh38
rs373444133 Chr.9: 99004846-99004846 on Build GRCh38
rs573694574 Chr.9: 99004875-99004875 on Build GRCh38
rs147872746 Chr.9: 99004995-99004995 on Build GRCh38
rs779538138 Chr.9: 99004927-99004927 on Build GRCh38
rs761575477 Chr.9: 99004955-99004955 on Build GRCh38
rs754398982 Chr.9: 99004922-99004922 on Build GRCh38
rs1017399668 Chr.9: 99005024-99005024 on Build GRCh38
rs759670653 Chr.9: 99004964-99004964 on Build GRCh38
rs905281294 Chr.9: 99004973-99004973 on Build GRCh38
rs761174334 Chr.9: 99004901-99004901 on Build GRCh38
rs766890603 Chr.9: 99005053-99005053 on Build GRCh38
rs757662358 Chr.9: 99004986-99004986 on Build GRCh38
rs768244834 Chr.9: 99005005-99005005 on Build GRCh38
rs561432064 Chr.9: 99004874-99004874 on Build GRCh38
rs779395704 Chr.9: 99004996-99004996 on Build GRCh38
rs147593729 Chr.9: 99004873-99004873 on Build GRCh38
rs373475447 Chr.9: 99004945-99004945 on Build GRCh38
rs764540735 Chr.9: 99004982-99004982 on Build GRCh38
rs756514581 Chr.9: 99004917-99004917 on Build GRCh38
rs760766648 Chr.9: 99004976-99004976 on Build GRCh38
rs776243720 Chr.9: 99004954-99004954 on Build GRCh38
rs929232672 Chr.9: 99004882-99004882 on Build GRCh38
rs35934703 Chr.9: 99005033-99005033 on Build GRCh38
rs954004407 Chr.9: 99004992-99004992 on Build GRCh38
rs761218857 Chr.9: 99004978-99004978 on Build GRCh38
rs771700078 Chr.9: 99004934-99004934 on Build GRCh38
rs910284041 Chr.9: 99004843-99004843 on Build GRCh38
rs148479358 Chr.9: 99004946-99004946 on Build GRCh38
rs763743004 Chr.9: 99004867-99004867 on Build GRCh38
rs757786955 Chr.9: 99004924-99004924 on Build GRCh38
rs767794809 Chr.9: 99004896-99004896 on Build GRCh38
rs752968243 Chr.9: 99004968-99004968 on Build GRCh38
rs752851060 Chr.9: 99004916-99004916 on Build GRCh38
rs751071936 Chr.9: 99005059-99005059 on Build GRCh38
rs1047612913 Chr.9: 99004925-99004925 on Build GRCh38
rs913699590 Chr.9: 99005015-99005015 on Build GRCh38
rs201273741 Chr.9: 99005035-99005035 on Build GRCh38
rs767526994 Chr.9: 99004965-99004965 on Build GRCh38
rs770465068 Chr.9: 99004860-99004861 on Build GRCh38
rs374155210 Chr.9: 99005066-99005066 on Build GRCh38
rs769764905 Chr.9: 99004962-99004962 on Build GRCh38
rs777931154 Chr.9: 99004920-99004920 on Build GRCh38
rs147720546 Chr.9: 99005032-99005032 on Build GRCh38
rs370219945 Chr.9: 99004997-99004997 on Build GRCh38
rs371128609 Chr.9: 99004871-99004871 on Build GRCh38
rs780868682 Chr.9: 99005008-99005008 on Build GRCh38
rs34138827 Chr.9: 99004963-99004963 on Build GRCh38
rs769564877 Chr.9: 99005020-99005020 on Build GRCh38
rs145422418 Chr.9: 99005029-99005029 on Build GRCh38
rs572154767 Chr.9: 99004864-99004864 on Build GRCh38
rs139529940 Chr.9: 99005052-99005052 on Build GRCh38
rs16918128 Chr.9: 99005021-99005021 on Build GRCh38
rs765633564 Chr.9: 99005036-99005036 on Build GRCh38
rs780768048 Chr.9: 99005063-99005063 on Build GRCh38
rs748005108 Chr.9: 99005017-99005017 on Build GRCh38
rs779774122 Chr.9: 99004940-99004940 on Build GRCh38
rs375676063 Chr.9: 99004859-99004859 on Build GRCh38
rs779626166 Chr.9: 99004988-99004988 on Build GRCh38
rs983796384 Chr.9: 99005047-99005047 on Build GRCh38
rs761619253 Chr.9: 99004872-99004872 on Build GRCh38
rs762809677 Chr.9: 99005026-99005026 on Build GRCh38
rs976322636 Chr.9: 99005013-99005013 on Build GRCh38
rs745407392 Chr.9: 99004932-99004932 on Build GRCh38
rs750948894 Chr.9: 99004992-99004992 on Build GRCh38
rs754507271 Chr.9: 99005060-99005060 on Build GRCh38

View Full Product Details