Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107906142-107906407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLD
Entrez Gene ID
1738
Gene Name
dihydrolipoamide dehydrogenase
Gene Aliases
DLDD, DLDH, E3, GCSL, LAD, PHE3
UniGene
Hs.131711
Gene Chromosome Location
Chr.7: 107891107-107921198 on Build GRCh38
Transcripts
NM_000108.4 NM_001289750.1 NM_001289751.1 NM_001289752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6175705 c.614G>C p.G205A Chr.7: 107906298-107906298 on Build GRCh38
4153745 c.670A>G p.I224V Chr.7: 107906354-107906354 on Build GRCh38
3255016 c.626_627insA p.V212fs*32 Chr.7: 107906310-107906311 on Build GRCh38
1447179 c.627delA p.V212fs*12 Chr.7: 107906311-107906311 on Build GRCh38
334750 c.659G>T p.G220V Chr.7: 107906343-107906343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764478905 Chr.7: 107906353-107906353 on Build GRCh38
rs772664162 Chr.7: 107906315-107906315 on Build GRCh38
rs200736488 Chr.7: 107906325-107906325 on Build GRCh38
rs1040811473 Chr.7: 107906310-107906311 on Build GRCh38
rs113165651 Chr.7: 107906188-107906188 on Build GRCh38
rs751374506 Chr.7: 107906372-107906372 on Build GRCh38
rs750449027 Chr.7: 107906361-107906361 on Build GRCh38
rs991936102 Chr.7: 107906224-107906224 on Build GRCh38
rs144590106 Chr.7: 107906293-107906293 on Build GRCh38
rs947296124 Chr.7: 107906250-107906251 on Build GRCh38
rs112693464 Chr.7: 107906196-107906196 on Build GRCh38
rs186031819 Chr.7: 107906264-107906264 on Build GRCh38
rs761089387 Chr.7: 107906333-107906333 on Build GRCh38
rs200181899 Chr.7: 107906324-107906324 on Build GRCh38
rs568211167 Chr.7: 107906360-107906360 on Build GRCh38
rs780025714 Chr.7: 107906369-107906369 on Build GRCh38
rs958733642 Chr.7: 107906221-107906221 on Build GRCh38
rs762002359 Chr.7: 107906200-107906200 on Build GRCh38
rs540793307 Chr.7: 107906186-107906186 on Build GRCh38
rs768003092 Chr.7: 107906326-107906326 on Build GRCh38
rs73726765 Chr.7: 107906359-107906359 on Build GRCh38
rs1057517016 Chr.7: 107906317-107906318 on Build GRCh38
rs867138403 Chr.7: 107906321-107906321 on Build GRCh38
rs75123588 Chr.7: 107906375-107906375 on Build GRCh38
rs952404558 Chr.7: 107906177-107906177 on Build GRCh38
rs373730165 Chr.7: 107906283-107906283 on Build GRCh38
rs746757721 Chr.7: 107906272-107906272 on Build GRCh38
rs747693486 Chr.7: 107906284-107906284 on Build GRCh38
rs777659984 Chr.7: 107906211-107906211 on Build GRCh38
rs758280079 Chr.7: 107906365-107906365 on Build GRCh38
rs759995075 Chr.7: 107906318-107906318 on Build GRCh38
rs775732459 Chr.7: 107906330-107906330 on Build GRCh38
rs768248572 Chr.7: 107906281-107906281 on Build GRCh38
rs754081428 Chr.7: 107906354-107906354 on Build GRCh38
rs181089830 Chr.7: 107906232-107906232 on Build GRCh38
rs556951509 Chr.7: 107906179-107906179 on Build GRCh38

View Full Product Details