Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138853941-138854212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECT2L
Entrez Gene ID
345930
Gene Name
epithelial cell transforming 2 like
Gene Aliases
ARHGEF32, C6orf91, FBXO49, LFDH, dJ509I19.2, dJ509I19.3, dJ509I19.5
UniGene
Hs.660603
Gene Chromosome Location
Chr.6: 138796107-138904070 on Build GRCh38
Transcripts
NM_001077706.2 NM_001195037.2 XM_006715472.3 XM_011535795.2 XM_011535797.2 XM_017010828.1 XM_017010829.1 XM_017010830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5779410 c.1192G>A p.A398T Chr.6: 138854148-138854148 on Build GRCh38
3621124 c.1179C>T p.F393F Chr.6: 138854135-138854135 on Build GRCh38
3621123 c.1139A>G p.Y380C Chr.6: 138854095-138854095 on Build GRCh38
5902769 c.1165C>T p.H389Y Chr.6: 138854121-138854121 on Build GRCh38
7327747 c.1171G>C p.D391H Chr.6: 138854127-138854127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030735369 Chr.6: 138854095-138854095 on Build GRCh38
rs767478902 Chr.6: 138854011-138854012 on Build GRCh38
rs765499367 Chr.6: 138854131-138854131 on Build GRCh38
rs776034165 Chr.6: 138854106-138854106 on Build GRCh38
rs573437951 Chr.6: 138854123-138854123 on Build GRCh38
rs750438274 Chr.6: 138854043-138854043 on Build GRCh38
rs371347697 Chr.6: 138854181-138854181 on Build GRCh38
rs903484817 Chr.6: 138854078-138854078 on Build GRCh38
rs372903555 Chr.6: 138854124-138854124 on Build GRCh38
rs367782648 Chr.6: 138854136-138854136 on Build GRCh38
rs762397526 Chr.6: 138854098-138854098 on Build GRCh38
rs1048117123 Chr.6: 138854183-138854183 on Build GRCh38
rs139396585 Chr.6: 138854091-138854091 on Build GRCh38
rs757068628 Chr.6: 138853980-138853980 on Build GRCh38
rs191760485 Chr.6: 138853981-138853981 on Build GRCh38
rs146285909 Chr.6: 138854013-138854013 on Build GRCh38
rs977240861 Chr.6: 138854119-138854119 on Build GRCh38
rs769367602 Chr.6: 138854053-138854053 on Build GRCh38
rs887914896 Chr.6: 138853983-138853983 on Build GRCh38
rs938578760 Chr.6: 138854128-138854128 on Build GRCh38
rs915546712 Chr.6: 138854164-138854164 on Build GRCh38
rs781190401 Chr.6: 138854174-138854174 on Build GRCh38
rs1015099124 Chr.6: 138854012-138854012 on Build GRCh38
rs751496361 Chr.6: 138854142-138854142 on Build GRCh38
rs769595524 Chr.6: 138854182-138854182 on Build GRCh38
rs757613141 Chr.6: 138853994-138853994 on Build GRCh38
rs529620990 Chr.6: 138854139-138854139 on Build GRCh38
rs754935876 Chr.6: 138854155-138854155 on Build GRCh38
rs762274234 Chr.6: 138854129-138854129 on Build GRCh38
rs200690490 Chr.6: 138854115-138854115 on Build GRCh38
rs150276683 Chr.6: 138854019-138854019 on Build GRCh38
rs777364346 Chr.6: 138854185-138854185 on Build GRCh38
rs774035174 Chr.6: 138854115-138854115 on Build GRCh38
rs1012366578 Chr.6: 138854006-138854006 on Build GRCh38
rs780095123 Chr.6: 138854012-138854012 on Build GRCh38
rs561406616 Chr.6: 138854100-138854100 on Build GRCh38
rs201910297 Chr.6: 138854135-138854135 on Build GRCh38
rs750462721 Chr.6: 138854002-138854002 on Build GRCh38
rs184996835 Chr.6: 138854027-138854027 on Build GRCh38
rs149517821 Chr.6: 138854101-138854101 on Build GRCh38
rs375332969 Chr.6: 138854117-138854117 on Build GRCh38
rs1028946716 Chr.6: 138854038-138854038 on Build GRCh38
rs754066113 Chr.6: 138853986-138853986 on Build GRCh38
rs1044728958 Chr.6: 138854049-138854049 on Build GRCh38
rs758566165 Chr.6: 138854011-138854011 on Build GRCh38
rs748978043 Chr.6: 138854190-138854190 on Build GRCh38
rs768469146 Chr.6: 138854019-138854019 on Build GRCh38
rs377533630 Chr.6: 138854023-138854023 on Build GRCh38
rs779131533 Chr.6: 138854001-138854001 on Build GRCh38
rs587777937 Chr.6: 138854025-138854025 on Build GRCh38

View Full Product Details