Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51478411-51478662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM18
Entrez Gene ID
729767
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
Gene Aliases
-
UniGene
Hs.653111
Gene Chromosome Location
Chr.19: 51478543-51491261 on Build GRCh38
Transcripts
NM_001278392.1 XM_011527258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
713158 c.139G>C p.V47L Chr.19: 51478598-51478598 on Build GRCh38
1736754 c.22A>G p.T8A Chr.19: 51478481-51478481 on Build GRCh38
4080617 c.150G>A p.E50E Chr.19: 51478609-51478609 on Build GRCh38
3835516 c.130G>T p.E44* Chr.19: 51478589-51478589 on Build GRCh38
4827699 c.183C>G p.L61L Chr.19: 51478642-51478642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202002299 Chr.19: 51478550-51478550 on Build GRCh38
rs970113981 Chr.19: 51478543-51478543 on Build GRCh38
rs757571916 Chr.19: 51478613-51478613 on Build GRCh38
rs201348861 Chr.19: 51478567-51478567 on Build GRCh38
rs140323408 Chr.19: 51478538-51478538 on Build GRCh38
rs867860393 Chr.19: 51478463-51478463 on Build GRCh38
rs753645495 Chr.19: 51478494-51478494 on Build GRCh38
rs373977687 Chr.19: 51478592-51478592 on Build GRCh38
rs760699612 Chr.19: 51478469-51478469 on Build GRCh38
rs371923661 Chr.19: 51478616-51478616 on Build GRCh38
rs190636927 Chr.19: 51478561-51478561 on Build GRCh38
rs747044053 Chr.19: 51478630-51478630 on Build GRCh38
rs377744953 Chr.19: 51478520-51478520 on Build GRCh38
rs374609909 Chr.19: 51478605-51478605 on Build GRCh38
rs11878388 Chr.19: 51478499-51478499 on Build GRCh38
rs763978927 Chr.19: 51478476-51478476 on Build GRCh38
rs543408165 Chr.19: 51478519-51478519 on Build GRCh38
rs576830448 Chr.19: 51478508-51478508 on Build GRCh38
rs371839400 Chr.19: 51478583-51478583 on Build GRCh38
rs760718373 Chr.19: 51478552-51478552 on Build GRCh38
rs754318036 Chr.19: 51478611-51478611 on Build GRCh38
rs779284544 Chr.19: 51478615-51478615 on Build GRCh38
rs746114060 Chr.19: 51478549-51478549 on Build GRCh38
rs560556048 Chr.19: 51478460-51478460 on Build GRCh38
rs201244522 Chr.19: 51478545-51478545 on Build GRCh38
rs758805765 Chr.19: 51478517-51478517 on Build GRCh38
rs929993549 Chr.19: 51478532-51478532 on Build GRCh38
rs267605616 Chr.19: 51478628-51478628 on Build GRCh38
rs544804629 Chr.19: 51478587-51478587 on Build GRCh38
rs1012070675 Chr.19: 51478609-51478609 on Build GRCh38
rs764914625 Chr.19: 51478505-51478505 on Build GRCh38
rs375100911 Chr.19: 51478620-51478620 on Build GRCh38
rs532919221 Chr.19: 51478582-51478582 on Build GRCh38
rs373783072 Chr.19: 51478539-51478539 on Build GRCh38
rs775610944 Chr.19: 51478551-51478551 on Build GRCh38
rs76009806 Chr.19: 51478625-51478625 on Build GRCh38
rs762661755 Chr.19: 51478590-51478590 on Build GRCh38
rs758969501 Chr.19: 51478623-51478623 on Build GRCh38
rs759549779 Chr.19: 51478451-51478451 on Build GRCh38
rs192964426 Chr.19: 51478604-51478604 on Build GRCh38
rs764895584 Chr.19: 51478465-51478465 on Build GRCh38
rs866326044 Chr.19: 51478507-51478507 on Build GRCh38
rs778197339 Chr.19: 51478536-51478536 on Build GRCh38
rs1038829557 Chr.19: 51478471-51478471 on Build GRCh38

View Full Product Details