Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75922107-75922332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB17
Entrez Gene ID
127247
Gene Name
ankyrin repeat and SOCS box containing 17
Gene Aliases
Asb-17
UniGene
Hs.125423
Gene Chromosome Location
Chr.1: 75918873-75937625 on Build GRCh38
Transcripts
NM_080868.2 NR_026546.1 XM_006710345.3 XM_006710346.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
253135 c.529_530insA p.N177fs*21 Chr.1: 75922231-75922232 on Build GRCh38
4495227 c.449C>T p.P150L Chr.1: 75922312-75922312 on Build GRCh38
7293371 c.459T>C p.Y153Y Chr.1: 75922302-75922302 on Build GRCh38
7323122 c.574G>A p.V192I Chr.1: 75922187-75922187 on Build GRCh38
6201553 c.602C>T p.A201V Chr.1: 75922159-75922159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776064305 Chr.1: 75922169-75922169 on Build GRCh38
rs368584559 Chr.1: 75922168-75922168 on Build GRCh38
rs771938087 Chr.1: 75922230-75922230 on Build GRCh38
rs778698257 Chr.1: 75922224-75922224 on Build GRCh38
rs776333372 Chr.1: 75922239-75922239 on Build GRCh38
rs777498692 Chr.1: 75922231-75922231 on Build GRCh38
rs745965498 Chr.1: 75922143-75922143 on Build GRCh38
rs951106482 Chr.1: 75922225-75922225 on Build GRCh38
rs771955199 Chr.1: 75922151-75922151 on Build GRCh38
rs200439606 Chr.1: 75922277-75922277 on Build GRCh38
rs775828543 Chr.1: 75922249-75922249 on Build GRCh38
rs756281376 Chr.1: 75922213-75922213 on Build GRCh38
rs758956251 Chr.1: 75922243-75922243 on Build GRCh38
rs150025062 Chr.1: 75922145-75922145 on Build GRCh38
rs758668303 Chr.1: 75922179-75922179 on Build GRCh38
rs868213936 Chr.1: 75922148-75922148 on Build GRCh38
rs764718283 Chr.1: 75922260-75922260 on Build GRCh38
rs778161358 Chr.1: 75922183-75922186 on Build GRCh38
rs769395035 Chr.1: 75922248-75922248 on Build GRCh38
rs752002447 Chr.1: 75922197-75922197 on Build GRCh38
rs909602377 Chr.1: 75922289-75922289 on Build GRCh38
rs1006157087 Chr.1: 75922246-75922246 on Build GRCh38
rs141282823 Chr.1: 75922191-75922191 on Build GRCh38
rs368309117 Chr.1: 75922254-75922254 on Build GRCh38
rs748161750 Chr.1: 75922225-75922225 on Build GRCh38
rs774346181 Chr.1: 75922149-75922149 on Build GRCh38
rs762270167 Chr.1: 75922296-75922296 on Build GRCh38
rs747284151 Chr.1: 75922132-75922132 on Build GRCh38
rs750458762 Chr.1: 75922207-75922207 on Build GRCh38
rs749460467 Chr.1: 75922230-75922231 on Build GRCh38
rs201937211 Chr.1: 75922178-75922178 on Build GRCh38
rs757379813 Chr.1: 75922136-75922136 on Build GRCh38
rs750711412 Chr.1: 75922305-75922305 on Build GRCh38
rs765658926 Chr.1: 75922173-75922173 on Build GRCh38
rs757504254 Chr.1: 75922198-75922198 on Build GRCh38
rs756120840 Chr.1: 75922310-75922310 on Build GRCh38
rs964711557 Chr.1: 75922308-75922308 on Build GRCh38
rs753305541 Chr.1: 75922177-75922177 on Build GRCh38
rs759092475 Chr.1: 75922170-75922170 on Build GRCh38
rs770506591 Chr.1: 75922235-75922235 on Build GRCh38
rs375630579 Chr.1: 75922206-75922206 on Build GRCh38
rs756221723 Chr.1: 75922136-75922136 on Build GRCh38
rs963926024 Chr.1: 75922301-75922301 on Build GRCh38
rs568802635 Chr.1: 75922138-75922138 on Build GRCh38
rs746821649 Chr.1: 75922233-75922233 on Build GRCh38
rs760283849 Chr.1: 75922164-75922164 on Build GRCh38
rs772823773 Chr.1: 75922155-75922155 on Build GRCh38
rs1016240958 Chr.1: 75922295-75922295 on Build GRCh38
rs767844980 Chr.1: 75922297-75922297 on Build GRCh38
rs950670142 Chr.1: 75922244-75922244 on Build GRCh38

View Full Product Details