Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 96289104-96289351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK17
Entrez Gene ID
5128
Gene Name
cyclin dependent kinase 17
Gene Aliases
PCTAIRE2, PCTK2
UniGene
Hs.506415
Gene Chromosome Location
Chr.12: 96278261-96400588 on Build GRCh38
Transcripts
NM_001170464.2 NM_002595.4 XM_017019405.1 XM_017019406.1 XM_017019407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
239270 c.998-1G>A p.? Chr.12: 96289288-96289288 on Build GRCh38
4212147 c.1073A>G p.D358G Chr.12: 96289212-96289212 on Build GRCh38
696277 c.998-1G>T p.? Chr.12: 96289288-96289288 on Build GRCh38
3704471 c.1091C>A p.S364* Chr.12: 96289194-96289194 on Build GRCh38
7427218 c.1053A>T p.T351T Chr.12: 96289232-96289232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780782161 Chr.12: 96289189-96289189 on Build GRCh38
rs989956909 Chr.12: 96289137-96289137 on Build GRCh38
rs760790261 Chr.12: 96289285-96289285 on Build GRCh38
rs945761693 Chr.12: 96289229-96289229 on Build GRCh38
rs754551308 Chr.12: 96289178-96289178 on Build GRCh38
rs753288277 Chr.12: 96289191-96289191 on Build GRCh38
rs769661692 Chr.12: 96289297-96289297 on Build GRCh38
rs563225357 Chr.12: 96289172-96289172 on Build GRCh38
rs773200145 Chr.12: 96289298-96289298 on Build GRCh38
rs759225376 Chr.12: 96289320-96289320 on Build GRCh38
rs762714806 Chr.12: 96289301-96289301 on Build GRCh38
rs746029909 Chr.12: 96289223-96289223 on Build GRCh38
rs369822114 Chr.12: 96289194-96289194 on Build GRCh38
rs775502433 Chr.12: 96289261-96289261 on Build GRCh38
rs779161958 Chr.12: 96289222-96289222 on Build GRCh38
rs766016499 Chr.12: 96289138-96289138 on Build GRCh38
rs934245081 Chr.12: 96289311-96289311 on Build GRCh38
rs201520694 Chr.12: 96289305-96289305 on Build GRCh38
rs914300756 Chr.12: 96289221-96289221 on Build GRCh38
rs530451291 Chr.12: 96289219-96289219 on Build GRCh38
rs544941188 Chr.12: 96289160-96289160 on Build GRCh38
rs766221371 Chr.12: 96289316-96289316 on Build GRCh38
rs369424893 Chr.12: 96289136-96289136 on Build GRCh38
rs372999926 Chr.12: 96289319-96289319 on Build GRCh38
rs749653666 Chr.12: 96289208-96289208 on Build GRCh38
rs45574339 Chr.12: 96289232-96289232 on Build GRCh38
rs200350039 Chr.12: 96289205-96289205 on Build GRCh38

View Full Product Details